Search
Search

Wnt11| Gene Symbol: Wnt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285792.1 | NP_001272721.1 | 4 - 5 | 816 | 95 | |
| NM_001285794.1 | NP_001272723.1 | 3 - 4 | 574 | 95 | ||
| NM_001285795.1 | NP_001272724.1 | 2 - 3 | 582 | 95 | ||
| NM_009519.2 | NP_033545.1 | 2 - 3 | 582 | 95 | ||
| GenBank mRNA | AK031573.1 | - | 2 - 3 | 425 | 95 | |
| AK077828.1 | - | 3 - 4 | 512 | 95 | ||
| AK138967.1 | - | 3 - 4 | 575 | 95 | ||
| BC125482.1 | - | 2 - 3 | 529 | 40129997 | 95 | |
| BC125484.1 | - | 2 - 3 | 529 | 40129999 | 95 | |
| BC144947.1 | - | 2 - 3 | 529 | 95 | ||
| X70800.1 | - | 2 - 3 | 589 | 95 | ||
Set Membership: |
|