Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00437347_m1
          See other WNT5A GE Assays ›
          Gene Symbol:
          Wnt5a
          Gene Name:
          wingless-type MMTV integration site family, member 5A
          Gene Aliases:
          8030457G12Rik, Wnt-5a
          Chromosome Location:
          Chr.14: 28505473 - 28527448 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00437347_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wnt5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256224.1 NP_001243153.1 4 - 5 695 70
          NM_009524.3 NP_033550.2 4 - 5 1318 70
          GenBank mRNA AK031512.1 - 4 - 5 1028 70
          AK031597.1 - 4 - 5 1291 70
          AK036824.1 - 4 - 5 696 70
          BC018425.1 - 4 - 5 935 3487288 70
          M89798.1 - 4 - 5 1229 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000290.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Wnt Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wnt5a

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          ameboidal-type cell migration
          establishment of planar polarity
          somitogenesis
          positive regulation of cytokine production
          neural tube closure
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          heart looping
          morphogenesis of an epithelium
          positive regulation of mesenchymal cell proliferation
          positive regulation of cytokine secretion involved in immune response
          type B pancreatic cell development
          pericardium morphogenesis
          axis elongation
          protein phosphorylation
          signal transduction
          Wnt signaling pathway, calcium modulating pathway
          JNK cascade
          activation of JUN kinase activity
          cell-cell signaling
          multicellular organism development
          determination of left/right symmetry
          axon guidance
          hindgut morphogenesis
          midgut development
          positive regulation of cell proliferation
          male gonad development
          anterior/posterior axis specification, embryo
          embryo development
          organ morphogenesis
          anterior/posterior pattern specification
          positive regulation of endothelial cell migration
          positive regulation of peptidyl-threonine phosphorylation
          positive regulation of T cell chemotaxis
          positive regulation of neuron projection development
          Wnt signaling pathway
          cell migration
          olfactory bulb interneuron development
          neural tube development
          positive regulation of cell-cell adhesion
          cell differentiation
          lung development
          embryonic limb morphogenesis
          negative regulation of BMP signaling pathway
          positive regulation of cGMP metabolic process
          activation of protein kinase B activity
          positive regulation of interferon-gamma production
          positive regulation of interleukin-6 production
          regulation of protein localization
          positive regulation of peptidyl-serine phosphorylation
          cellular protein localization
          limb morphogenesis
          non-canonical Wnt signaling pathway
          post-anal tail morphogenesis
          non-canonical Wnt signaling pathway via JNK cascade
          negative regulation of fibroblast growth factor receptor signaling pathway
          wound healing
          inner ear morphogenesis
          embryonic digit morphogenesis
          positive regulation of macrophage activation
          negative regulation of apoptotic process
          positive regulation of JUN kinase activity
          positive regulation of chemokine biosynthetic process
          cell fate commitment
          negative regulation of fat cell differentiation
          positive regulation of protein catabolic process
          positive regulation of angiogenesis
          positive regulation of ossification
          positive regulation of endocytosis
          positive regulation of meiotic nuclear division
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          development of primary male sexual characteristics
          positive regulation of fibroblast proliferation
          digestive tract morphogenesis
          embryonic skeletal system development
          genitalia development
          neuron projection morphogenesis
          negative regulation of axon extension involved in axon guidance
          hypophysis morphogenesis
          positive regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          positive regulation of interleukin-1 beta secretion
          positive regulation of inflammatory response
          negative chemotaxis
          positive regulation of NF-kappaB transcription factor activity
          cartilage development
          negative regulation of synapse assembly
          palate development
          convergent extension
          convergent extension involved in organogenesis
          uterus development
          cervix development
          vagina development
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          urinary bladder development
          face development
          positive regulation of type I interferon-mediated signaling pathway
          lateral sprouting involved in mammary gland duct morphogenesis
          tube closure
          mesenchymal-epithelial cell signaling
          negative regulation of prostatic bud formation
          mammary gland branching involved in thelarche
          epithelial cell proliferation involved in mammary gland duct elongation
          positive regulation of response to cytokine stimulus
          regulation of branching involved in mammary gland duct morphogenesis
          positive regulation of macrophage cytokine production
          positive regulation of cartilage development
          planar cell polarity pathway involved in outflow tract morphogenesis
          planar cell polarity pathway involved in ventricular septum morphogenesis
          planar cell polarity pathway involved in cardiac right atrium morphogenesis
          planar cell polarity pathway involved in cardiac muscle tissue morphogenesis
          planar cell polarity pathway involved in pericardium morphogenesis
          positive regulation of thymocyte apoptotic process
          cellular response to molecule of bacterial origin
          hematopoietic stem cell proliferation
          dopaminergic neuron differentiation
          negative regulation of mesenchymal cell proliferation
          primitive streak formation
          positive regulation of protein kinase C signaling
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          planar cell polarity pathway involved in neural tube closure
          positive regulation of protein kinase C activity
          positive regulation of cell-cell adhesion mediated by cadherin
          positive regulation of interleukin-8 secretion
          transcription factor activity, sequence-specific DNA binding
          receptor binding
          frizzled binding
          frizzled-2 binding
          receptor tyrosine kinase-like orphan receptor binding
          cytokine activity
          protein binding
          protein domain specific binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400002353.mm.1 NM_001256224.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002353.mm.1 NM_001256224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002353.mm.1 NM_009524.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002353.mm.1 NM_009524.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002353.mm.1 TC1400000290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002354.mm.1 NM_001256224.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002354.mm.1 NM_001256224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002354.mm.1 NM_009524.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002354.mm.1 NM_009524.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002354.mm.1 TC1400000290.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400002352.mm.1 NM_001256224.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002352.mm.1 NM_001256224.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002352.mm.1 NM_009524.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002352.mm.1 NM_009524.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400002352.mm.1 TC1400000290.mm.1 MTA TC

          Back To Top
           

          Citations (1)

          Wnt10b deficiency promotes coexpression of myogenic and adipogenic programs in myoblasts.
          Authors
          Vertino AM, Taylor-Jones JM, Longo KA, Bearden ED, Lane TF, McGehee RE Jr, MacDougald OA, Peterson CA
          Journal
          Mol Biol Cell - Page No 2039 (Fri Apr 01 00:00:00 UTC 2005)
          Volume (Year)
          16 (2005)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline