Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00437657_m1
          See other ATP4B GE Assays ›
          Gene Symbol:
          Atp4b
          Gene Name:
          ATPase, H+/K+ exchanging, beta polypeptide
          Gene Aliases:
          AV080843
          Chromosome Location:
          Chr.8: 13386209 - 13396778 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00437657_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009724.2 NP_033854.1 5 - 6 637 125
          GenBank mRNA AK008180.1 - 5 - 6 636 125
          AK009499.1 - 5 - 6 637 125
          AK009670.1 - 5 - 6 642 124
          AK009994.1 - 5 - 6 636 125
          AK010003.1 - 5 - 6 636 125
          AK010049.1 - 5 - 6 684 125
          BC120705.1 - 5 - 6 636 40129628 125
          BC125292.1 - 5 - 6 636 40129807 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001736.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp4b

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          potassium ion transport
          sodium ion transport
          cell adhesion
          regulation of proton transport
          proton transport
          protein glycosylation in endoplasmic reticulum
          pH reduction
          potassium ion transmembrane transport
          hydrogen ion transmembrane transport
          hydrogen:potassium-exchanging ATPase activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959909 AK010003.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959909 AK010030.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959909 AK010049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959909 AK009499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959909 AK009732.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451948 AK041828.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746252 AB232613.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746252 AK017590.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746252 BC032936.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746252 NM_011226.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746252 X80473.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 106024 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119673 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104794 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143795 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959909 AK010003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959909 AK010030.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959909 AK010049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959909 AK009499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959909 AK009732.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P319180 AK010003.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P319180 AK010030.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P319180 AK010049.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P319180 AK009499.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P319180 AK009732.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25667999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25667997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25668050 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104794 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119673 - -
          Whole Mouse Genome G4122A A_52_P319180 AK010003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319180 AK010030.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319180 AK010049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319180 AK009499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P319180 AK009732.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128097 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101073 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119673 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104794 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143795 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07433327 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07433329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06808131 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06960954 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07433326 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06739480 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06960962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06739477 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06960963 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06739470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06808136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25667975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25668023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06799688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06838971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06960956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06960961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06960953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06960939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25667994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25668015 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577158 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577155 - -
          Affymetrix HT MG-430 PM Array Plate 1448911_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577154 - -
          Mouse Genome 430 2.0 Array 1448911_PM_at - -
          Mouse Genome 430 2.0 Array 1448911_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11945_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11945_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800014036.mm.1 NM_009724.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014036.mm.1 NM_009724.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014036.mm.1 TC0800001736.mm.1 MTA TC
          Mouse Expression Set 430 1448911_at - -
          Mouse Expression Set 430 1448911_PM_at - -
          Murine Genome U74v2 Set 100501_PM_at - -
          Murine Genome U74v2 Set 100501_at - -
          Mouse Genome 430A 2.0 Array 1448911_PM_at - -
          Mouse Genome 430A 2.0 Array 1448911_at - -
          Affymetrix HT Mouse Genome 430A Array 1448911_at - -
          Affymetrix HT Mouse Genome 430A Array 1448911_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 802387 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1740156 - -
          Illumina mouse-6 v1.0 expression beadchip 1740156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2703480 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2703480 - -
          mouse-6 v1.1 (Illumina) scl35077.7.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35077.7.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2703480 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2703480 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline