Search
Search

Ccna2| Gene Symbol: Ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009828.2 | NP_033958.2 | 1 - 2 | 362 | 83 | |
| GenBank mRNA | AK044924.1 | - | 1 - 2 | 365 | 83 | |
| AK135861.1 | - | 1 - 2 | 334 | 83 | ||
| AK188335.1 | - | 1 - 2 | 29 | 83 | ||
| AK216847.1 | - | 1 - 2 | 34 | 83 | ||
| BB642910.1 | - | 1 - 2 | 362 | 83 | ||
| BC052730.1 | - | 1 - 2 | 310 | 6816226 | 83 | |
| X75483.1 | - | 1 - 2 | 310 | 83 | ||
| Z26580.1 | - | 1 - 2 | 319 | 83 | ||
Set Membership: |
|