| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036769 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036768 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036751 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036767 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036775 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036770 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036782 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036781 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036750 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036779 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036776 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036772 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036777 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036778 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036752 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036771 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036780 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2122987 |
BC058852.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
31715 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371090 |
AK077092.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371090 |
BC050188.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371090 |
NM_177742.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
956096 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
129937 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135313 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2122987 |
BC058852.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P503466 |
U79738.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P503466 |
BC058852.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P503466 |
NM_010055.3 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036768 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036785 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036764 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036769 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036743 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036751 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036746 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036741 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036766 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036759 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036780 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036736 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036754 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036749 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036740 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036789 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036763 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036775 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036760 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036782 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036750 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036777 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036773 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036774 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036742 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036767 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036757 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036786 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036747 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036738 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036735 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036772 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036745 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036771 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28036783 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135313 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P503466 |
U79738.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P503466 |
BC058852.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P503466 |
NM_010055.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104529 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135313 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09628071 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09495019 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09495027 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09410088 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09410097 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036780 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036768 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036775 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036778 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036779 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036767 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036752 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036769 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036751 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036770 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036776 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036772 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036781 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036771 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036777 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036750 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28036782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036773 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036729 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036751 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036763 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036849 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036851 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036768 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036870 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036856 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036783 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036842 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036754 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036741 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036860 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036818 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036760 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036766 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036750 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036834 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036757 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036727 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036806 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036802 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036815 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036714 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036716 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036771 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036819 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036764 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036725 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036825 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036844 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036840 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036866 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036767 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036812 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036722 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036719 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036800 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036831 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036824 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036774 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036772 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036795 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036775 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036817 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036837 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036871 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036797 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036743 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036739 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036742 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036780 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28036790 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036740 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036775 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036749 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036767 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036773 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036746 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036739 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036783 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036760 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036854 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036791 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036827 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09101544 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036754 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09703497 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036725 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036853 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036743 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036840 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036759 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036824 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036814 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09473190 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036771 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09465164 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036831 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09093346 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09171040 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036837 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036764 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036817 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036807 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036844 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036747 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09465155 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09644522 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036757 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036846 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036734 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09473188 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036806 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036729 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036774 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036819 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036772 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036821 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09465166 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036849 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036812 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036794 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036786 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09473194 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036842 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036738 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036852 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036785 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09093356 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036726 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036804 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09703499 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036763 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036809 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036828 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036836 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036736 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036752 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09101545 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09644520 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09473196 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09520405 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036818 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09465168 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036750 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036728 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036732 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036766 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036851 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036733 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P28036780 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P09473192 |
- |
- |