Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00438463_m1
          See other DNASE2A GE Assays ›
          Gene Symbol:
          Dnase2a
          Gene Name:
          deoxyribonuclease II alpha
          Gene Aliases:
          Dnase2
          Chromosome Location:
          Chr.8: 84908624 - 84911461 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00438463_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnase2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010062.3 NP_034192.1 5 - 6 874 71
          GenBank mRNA AK145192.1 - 5 - 6 875 71
          AK167277.1 - 5 - 6 840 71
          BC058609.1 - 5 - 6 821 5397016 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000972.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnase2a

          Panther Classification:

          Molecular Function -

          endodeoxyribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA catabolic process, endonucleolytic
          DNA metabolic process
          DNA catabolic process
          apoptotic DNA fragmentation
          apoptotic process
          multicellular organism development
          erythrocyte differentiation
          nucleic acid phosphodiester bond hydrolysis
          nuclease activity
          endonuclease activity
          endodeoxyribonuclease activity
          deoxyribonuclease II activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022589 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022585 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022588 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022583 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022584 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022587 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173005 BC060661.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173005 BY757082.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173005 NM_010062.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2173005 AK167277.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746321 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118068 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173005 BC060661.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173005 BY757082.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173005 NM_010062.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2173005 AK167277.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P453508 BC060661.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P453508 AK010630.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P380129 AK018651.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022596 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118068 - -
          Whole Mouse Genome G4122A A_52_P453508 BC060661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453508 AK010630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P380129 AK018651.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136287 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113154 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107290 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118068 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07309111 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07153742 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07309126 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07309115 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022585 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022588 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022589 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022587 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022583 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07241561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07153742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06716780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022598 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10573471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573463 - -
          Affymetrix HT MG-430 PM Array Plate 1448986_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1430135_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10573461 - -
          Mouse Genome 430 2.0 Array 1430135_PM_at - -
          Mouse Genome 430 2.0 Array 1448986_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448986_x_at - -
          Mouse Genome 430 2.0 Array 1430135_at - -
          Affymetrix Mouse Clariom S Array PSR0800007353.mm.1 NM_010062.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007353.mm.1 NM_010062.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007353.mm.1 TC0800000972.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007355.mm.1 NM_010062.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007355.mm.1 NM_010062.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007355.mm.1 TC0800000972.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007356.mm.1 NM_010062.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007356.mm.1 NM_010062.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007356.mm.1 TC0800000972.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007358.mm.1 NM_010062.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007358.mm.1 NM_010062.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007358.mm.1 TC0800000972.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007350.mm.1 NM_010062.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007350.mm.1 NM_010062.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007350.mm.1 TC0800000972.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007352.mm.1 NM_010062.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007352.mm.1 NM_010062.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007352.mm.1 TC0800000972.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007354.mm.1 NM_010062.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007354.mm.1 NM_010062.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007354.mm.1 TC0800000972.mm.1 MTA TC
          Mouse Expression Set 430 1448986_x_at - -
          Mouse Expression Set 430 1448986_PM_x_at - -
          Mouse Expression Set 430 1430135_PM_at - -
          Mouse Expression Set 430 1430135_at - -
          Mouse Genome 430A 2.0 Array 1448986_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1448986_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1430135_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430135_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 872680 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105700373 - -
          MouseWG-6 v2 Expression BeadChip 7050576 - -
          Illumina mouse-6 v1.0 expression beadchip 105700373 - -
          Illumina mouse-6 v1.0 expression beadchip 7050576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771000 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2435394 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771000 - -
          mouse-6 v1.1 (Illumina) scl33568.4.1_64-S - -
          mouse-6 v1.1 (Illumina) scl013423.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 105700373 - -
          Illumina Sentrix mouse-6.v1 bead array 7050576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013423.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050576 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105700373 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2975892 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2435392 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2627328 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2975892 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline