Search
Search

Eif2ak3| Gene Symbol: Eif2ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010121.2 | NP_034251.2 | 4 - 5 | 954 | 62 | |
| GenBank mRNA | AF076681.1 | - | 4 - 5 | 946 | 62 | |
| AK139679.1 | - | 2 - 3 | 276 | 62 | ||
| BC054809.1 | - | 4 - 5 | 966 | 6839221 | 62 | |
Set Membership: |
|