Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439403_m1
          See other IAPP GE Assays ›
          Gene Symbol:
          Iapp
          Gene Name:
          islet amyloid polypeptide
          Gene Aliases:
          DAP
          Chromosome Location:
          Chr.6: 142298425 - 142303820 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439403_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Iapp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010491.2 NP_034621.1 2 - 3 177 67
          GenBank mRNA BC027527.1 - 2 - 3 148 1379049 67
          CJ042995.1 - 2 - 3 180 67
          M25389.1 - 1 - 2 84 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001677.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Stem Cell Pluripotency 1700 Inventoried Mouse_Stem_Cell Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Iapp

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception of pain
          eating behavior
          negative regulation of cell differentiation
          negative regulation of bone resorption
          hormone activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114856 M25389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114856 NM_010491.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114856 BC027527.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178726 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114856 M25389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114856 NM_010491.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114856 BC027527.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P119841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24903758 - -
          Whole Mouse Genome G4122A A_51_P119841 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114856 M25389.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114856 NM_010491.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114856 BC027527.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006436 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05828302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006423 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05006444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24903736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05322727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05379344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05322728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05006435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24903724 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542633 - -
          Affymetrix HT MG-430 PM Array Plate 1423509_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1437323_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1456004_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1423510_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10542632 - -
          Mouse Genome 430 2.0 Array 1423509_PM_a_at - -
          Mouse Genome 430 2.0 Array 1437323_a_at - -
          Mouse Genome 430 2.0 Array 1423510_PM_at - -
          Mouse Genome 430 2.0 Array 1437323_PM_a_at - -
          Mouse Genome 430 2.0 Array 1456004_PM_x_at - -
          Mouse Genome 430 2.0 Array 1423510_at - -
          Mouse Genome 430 2.0 Array 1456004_x_at - -
          Mouse Genome 430 2.0 Array 1423509_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15874_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15874_at - -
          Affymetrix Mouse Clariom S Array PSR0600013358.mm.1 NM_010491.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013358.mm.1 NM_010491.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600013358.mm.1 TC0600001677.mm.1 MTA TC
          Mouse Expression Set 430 1423509_a_at - -
          Mouse Expression Set 430 1423510_at - -
          Mouse Expression Set 430 1423509_PM_a_at - -
          Mouse Expression Set 430 1423510_PM_at - -
          Mouse Expression Set 430 1456004_PM_x_at - -
          Mouse Expression Set 430 1456004_x_at - -
          Mouse Expression Set 430 1437323_PM_a_at - -
          Mouse Expression Set 430 1437323_a_at - -
          Murine Genome U74v2 Set 99488_PM_at - -
          Murine Genome U74v2 Set 99488_at - -
          Mouse Genome 430A 2.0 Array 1423509_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1456004_x_at - -
          Mouse Genome 430A 2.0 Array 1437323_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423510_PM_at - -
          Mouse Genome 430A 2.0 Array 1456004_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437323_a_at - -
          Mouse Genome 430A 2.0 Array 1423509_a_at - -
          Mouse Genome 430A 2.0 Array 1423510_at - -
          Affymetrix HT Mouse Genome 430A Array 1437323_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423510_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423509_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437323_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1456004_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1456004_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1423510_at - -
          Affymetrix HT Mouse Genome 430A Array 1423509_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 865893 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230040 - -
          Illumina mouse-6 v1.0 expression beadchip 4230040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248774 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248774 - -
          mouse-6 v1.1 (Illumina) scl29386.3.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230040 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29386.3.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230040 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2996479 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2996479 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline