Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439552_s1
          See other IFNB1 GE Assays ›
          Gene Symbol:
          Ifnb1
          Gene Name:
          interferon beta 1, fibroblast
          Gene Aliases:
          IFN-beta, IFNB, Ifb
          Chromosome Location:
          Chr.4: 88522025 - 88522794 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439552_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifnb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010510.1 NP_034640.1 1 638 69
          GenBank mRNA K00020.1 - 1 638 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003001.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifnb1

          Panther Classification:

          Molecular Function -

          interferon superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          adaptive immune response
          T cell activation involved in immune response
          B cell activation involved in immune response
          natural killer cell activation involved in immune response
          defense response
          humoral immune response
          cytokine-mediated signaling pathway
          B cell differentiation
          positive regulation of peptidyl-serine phosphorylation of STAT protein
          cellular response to interferon-beta
          B cell proliferation
          defense response to bacterium
          response to exogenous dsRNA
          negative regulation of viral genome replication
          negative regulation of T cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          defense response to virus
          type I interferon signaling pathway
          cellular response to dexamethasone stimulus
          negative regulation of T-helper 2 cell cytokine production
          positive regulation of apoptotic signaling pathway
          cytokine activity
          cytokine receptor binding
          type I interferon receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144180 K00020.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144180 NM_010510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144180 K00020.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144180 NM_010510.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164364 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144180 K00020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144180 NM_010510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144180 K00020.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144180 NM_010510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144180 K00020.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144180 NM_010510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144180 K00020.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144180 NM_010510.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23090989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23090994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23090988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23090998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23090990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23090992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23090996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091044 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164364 - -
          Whole Mouse Genome G4122A A_51_P144180 K00020.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144180 NM_010510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144180 K00020.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144180 NM_010510.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101906 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03942607 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03942603 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03942617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03942614 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03942619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03942613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23090957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090969 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03777788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03792644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03613291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03924754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03649141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03924749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03777791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03942613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03793312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03649138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23090976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091027 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514286 - -
          Affymetrix HT MG-430 PM Array Plate 1422305_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514285 - -
          Mouse Genome 430 2.0 Array 1422305_PM_at - -
          Mouse Genome 430 2.0 Array 1422305_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15977_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15977_at - -
          Affymetrix Mouse Clariom S Array PSR0400024291.mm.1 NM_010510.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024291.mm.1 NM_010510.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024291.mm.1 TC0400003001.mm.1 MTA TC
          Mouse Expression Set 430 1422305_PM_at - -
          Mouse Expression Set 430 1422305_at - -
          Murine Genome U74v2 Set 94145_at - -
          Murine Genome U74v2 Set 94145_PM_at - -
          Mouse Genome 430A 2.0 Array 1422305_at - -
          Mouse Genome 430A 2.0 Array 1422305_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422305_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422305_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 823832 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1400142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711910 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711910 - -
          mouse-6 v1.1 (Illumina) scl24132.1_469-S - -
          Illumina Sentrix mouse-6.v1 bead array 1400142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24132.1_469-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2711910 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2711910 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline