Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439564_m1
          See other IGF2 GE Assays ›
          Gene Symbol:
          Igf2
          Gene Name:
          insulin-like growth factor 2
          Gene Aliases:
          AL033362, Igf-2, Igf-II, M6pr, Mpr, Peg2
          Chromosome Location:
          Chr.7: 142650768 - 142662270 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439564_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001122736.1 NP_001116208.1 2 - 3 265 107
          NM_001122737.1 NP_001116209.1 2 - 3 255 107
          NM_010514.3 NP_034644.2 2 - 3 602 107
          GenBank mRNA BC053489.1 - 2 - 3 241 30013295 107
          BC058615.1 - 2 - 3 240 5686066 107
          CF742200.1 - 2 - 3 461 107
          CN460564.1 - 2 - 3 477 107
          M14951.1 - 2 - 3 186 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004571.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igf2

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          positive regulation of protein phosphorylation
          carbohydrate metabolic process
          glucose metabolic process
          memory
          organ morphogenesis
          response to organic cyclic compound
          exocrine pancreas development
          response to nicotine
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          positive regulation of activated T cell proliferation
          response to drug
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          positive regulation of glycogen biosynthetic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of natural killer cell mediated cytotoxicity
          positive regulation of insulin receptor signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          striated muscle cell differentiation
          positive regulation of cell division
          positive regulation of protein kinase B signaling
          positive regulation of protein serine/threonine kinase activity
          positive regulation of steroid hormone biosynthetic process
          positive regulation of receptor activity
          positive regulation of glycogen (starch) synthase activity
          insulin receptor binding
          insulin-like growth factor receptor binding
          hormone activity
          protein binding
          growth factor activity
          receptor activator activity
          protein serine/threonine kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700038552.mm.1 NM_001122736.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038552.mm.1 NM_001122737.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038552.mm.1 NM_010514.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038552.mm.1 NM_010514.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038552.mm.1 TC0700004571.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline