Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439620_m1
          See other IL1A GE Assays ›
          Gene Symbol:
          Il1a
          Gene Name:
          interleukin 1 alpha
          Gene Aliases:
          Il-1a
          Chromosome Location:
          Chr.2: 129299609 - 129310901 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439620_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010554.4 NP_034684.2 5 - 6 557 68
          GenBank mRNA AK156449.1 - 5 - 6 558 68
          BC003727.1 - 4 - 5 526 3599550 68
          M14639.1 - 1 - 2 191 68
          X01450.1 - 5 - 6 557 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004599.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Chemokine Panel Mouse Estrogen Panel Mouse Immune Response Mouse Alzheimer's Disease Inventoried Mouse_Immune_Response Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il1a

          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          fever generation
          connective tissue replacement involved in inflammatory response wound healing
          inflammatory response
          immune response
          negative regulation of cell proliferation
          positive regulation of vascular endothelial growth factor production
          positive regulation of steroid biosynthetic process
          cytokine-mediated signaling pathway
          keratinization
          positive regulation of prostaglandin secretion
          positive regulation of interleukin-6 production
          positive regulation of stress-activated MAPK cascade
          cellular response to heat
          ectopic germ cell programmed cell death
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of interleukin-2 biosynthetic process
          positive regulation of angiogenesis
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of JNK cascade
          response to copper ion
          positive regulation of cytokine secretion
          positive regulation of cell division
          regulation of sensory perception of pain
          positive regulation of ERK1 and ERK2 cascade
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of neutrophil migration
          cytokine activity
          interleukin-1 receptor binding
          copper ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200039399.mm.1 NM_010554.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039399.mm.1 NM_010554.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039399.mm.1 TC0200004599.mm.1 MTA TC

          Back To Top
           

          Citations (1)

          Accelerated recovery from acute hypoxia in obese mice is due to obesity-associated up-regulation of interleukin-1 receptor antagonist.
          Authors
          Sherry CL, Kim SS, Freund GG
          Journal
          Endocrinology - Page No 2660 (Mon Jun 01 00:00:00 UTC 2009)
          Volume (Year)
          150 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline