Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00439825_m1
          See other ITGB5 GE Assays ›
          Gene Symbol:
          Itgb5
          Gene Name:
          integrin beta 5
          Gene Aliases:
          AA475909, AI874634, ESTM23, [b]-5, [b]5, [b]5A, [b]5B, beta-5, beta5
          Chromosome Location:
          Chr.16: 33829665 - 33949338 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00439825_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgb5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145884.1 NP_001139356.1 1 - 2 369 96
          NM_010580.2 NP_034710.2 1 - 2 369 96
          GenBank mRNA AF022110.1 - 1 - 2 368 96
          AF043256.1 - 1 - 2 274 96
          AF043257.1 - 1 - 2 274 96
          AK087476.1 - 1 - 2 115 96
          BC046280.1 - 1 - 2 278 6394641 96
          BC058246.1 - 1 - 2 333 6313668 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000480.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgb5

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          cell-matrix adhesion
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          stress fiber assembly
          epithelial cell-cell adhesion
          receptor activity
          receptor binding
          integrin binding
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600004543.mm.1 NM_001145884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004543.mm.1 NM_001145884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004543.mm.1 NM_010580.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004543.mm.1 NM_010580.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004543.mm.1 TC1600000480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600004545.mm.1 NM_001145884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004545.mm.1 NM_001145884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004545.mm.1 NM_010580.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004545.mm.1 NM_010580.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004545.mm.1 TC1600000480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600004544.mm.1 NM_001145884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004544.mm.1 NM_001145884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004544.mm.1 NM_010580.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004544.mm.1 NM_010580.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004544.mm.1 TC1600000480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600004546.mm.1 NM_001145884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004546.mm.1 NM_001145884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004546.mm.1 NM_010580.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004546.mm.1 NM_010580.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600004546.mm.1 TC1600000480.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 604299 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline