Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440078_m1
          See other KCNK7 GE Assays ›
          Gene Symbol:
          Kcnk7
          Gene Name:
          potassium channel, subfamily K, member 7
          Gene Aliases:
          2310014G03Rik, Kcnk6, Kcnk8, Knot, Mlk3
          Chromosome Location:
          Chr.19: 5704476 - 5707405 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440078_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnk7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010609.2 NP_034739.2 1 - 2 315 82
          GenBank mRNA AB015729.1 - 1 - 2 360 82
          AF012324.1 - 1 - 2 375 82
          AF022820.1 - 1 - 2 312 82
          AF110521.1 - 1 - 2 426 82
          AK160180.1 - 1 - 2 322 82
          BC106993.2 - 1 - 2 315 40044182 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000080.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnk7

          Panther Classification:

          Molecular Function -

          ion channel transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          potassium ion transport
          stabilization of membrane potential
          regulation of ion transmembrane transport
          potassium ion transmembrane transport
          voltage-gated ion channel activity
          potassium channel activity
          potassium ion leak channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147692 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360435 AF022820.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P360435 AF022820.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P354018 AF110521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360435 AF022820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360435 AF022820.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P354018 AF110521.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108278 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460662 - -
          Affymetrix HT MG-430 PM Array Plate 1425437_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460658 - -
          Mouse Genome 430 2.0 Array 1425437_PM_a_at - -
          Mouse Genome 430 2.0 Array 1425437_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16530_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16530_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900000962.mm.1 NM_010609.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000962.mm.1 TC1900000080.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000963.mm.1 NM_010609.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000963.mm.1 TC1900000080.mm.1 MTA TC
          Mouse Expression Set 430 1425437_a_at - -
          Mouse Expression Set 430 1425437_PM_a_at - -
          Murine Genome U74v2 Set 95381_PM_at - -
          Murine Genome U74v2 Set 95381_at - -
          Mouse Genome 430A 2.0 Array 1425437_a_at - -
          Mouse Genome 430A 2.0 Array 1425437_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425437_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1425437_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 330112 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4230576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225334 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225334 - -
          mouse-6 v1.1 (Illumina) scl52852.2.1_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52852.2.1_67-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225334 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline