Search
Search
| Gene Symbol: Lta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010735.2 | NP_034865.1 | 2 - 3 | 303 | 59 | |
| GenBank mRNA | AK080308.1 | - | 2 - 3 | 166 | 59 | |
| AK153694.1 | - | 2 - 3 | 166 | 59 | ||
| AK156659.1 | - | 1 | 142 | |||
| BC099464.1 | - | 2 - 3 | 303 | 30846267 | 59 | |
| M16819.1 | - | 1 - 2 | 101 | 59 | ||
| X14800.1 | - | 1 - 2 | 87 | 59 | ||
Set Membership: |
|