| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582015 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581955 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581945 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581946 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581948 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581947 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581958 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582016 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582013 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581951 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581950 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581949 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581952 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581953 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581956 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582014 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581954 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002291 |
U62523.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002287 |
BC137578.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002287 |
U62523.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002287 |
AK012556.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002287 |
BC051983.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2002287 |
BC137577.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
AK003535.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
AK030532.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
AK042380.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
AK083365.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
BC020080.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
BC023802.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
NM_197986.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580381 |
NM_199145.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
582753 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P108854 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002287 |
BC137578.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002287 |
U62523.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002287 |
AK012556.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002287 |
BC051983.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002287 |
BC137577.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2002291 |
U62523.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P364302 |
U62523.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581953 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581978 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581990 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581945 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581984 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581952 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581975 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581991 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581941 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581964 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581937 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581982 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581949 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581946 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581963 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581981 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581956 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581993 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581960 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581957 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581985 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581973 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581977 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581971 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581974 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581951 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581988 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581940 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581938 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581962 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581948 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581983 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581954 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581943 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581961 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581965 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581935 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581955 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581992 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581958 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581979 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581969 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25581987 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P108854 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P364302 |
U62523.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665478 |
U62523.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665478 |
AK012556.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665478 |
BC051983.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P665478 |
NM_010836.3 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124161 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P108854 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06237455 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06237420 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06237435 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06237451 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06237457 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06237431 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06237442 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582013 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582015 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581954 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581953 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581951 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581952 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582014 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581956 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581949 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581950 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581946 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581948 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581945 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581947 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25582016 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581955 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25581958 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581907 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581897 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582007 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582003 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582010 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581973 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581998 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581971 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581975 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581990 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581902 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581949 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581964 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581958 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581883 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581877 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581978 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581969 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581869 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581960 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581986 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581895 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581888 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581894 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581955 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582015 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581963 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582018 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581892 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581983 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581874 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581885 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581872 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581867 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581899 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581953 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25582022 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581937 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581890 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581961 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581977 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581965 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25581948 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581946 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06237435 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581990 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581941 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582005 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06607186 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581960 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581953 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05989045 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05988002 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05987992 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581949 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581964 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582013 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581948 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581991 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581965 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05989043 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06237458 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581931 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582015 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582007 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582004 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581933 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581986 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581975 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582018 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06237451 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581995 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582020 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581979 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581973 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581977 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581988 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581940 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581978 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581958 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581937 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582017 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581935 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581969 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581962 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06237422 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581954 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581943 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05988012 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581945 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581985 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25582022 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581938 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581987 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581983 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581955 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05989041 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06237457 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25581971 |
- |
- |