Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440485_m1
          See other NOS2 GE Assays ›
          Gene Symbol:
          Nos2
          Gene Name:
          nitric oxide synthase 2, inducible
          Gene Aliases:
          NOS-II, Nos-2, Nos2a, i-NOS, iNOS
          Chromosome Location:
          Chr.11: 78920859 - 78960232 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440485_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (37)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nos2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010927.3 NP_035057.1 4 - 5 487 70
          GenBank mRNA AF065919.1 - 4 - 5 484 70
          AF065920.1 - 4 - 5 484 70
          AF065921.2 - 4 - 5 484 70
          AF065922.2 - 3 - 4 357 70
          AF065923.2 - 3 - 4 357 70
          AY090567.1 - 3 - 4 307 70
          BC062378.1 - 4 - 5 496 4978648 70
          M84373.1 - 4 - 5 488 70
          M87039.1 - 4 - 5 559 70
          M92649.1 - 4 - 5 495 70
          U43428.1 - 4 - 5 375 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001171.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nos2

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          ovulation from ovarian follicle
          response to hypoxia
          arginine catabolic process
          superoxide metabolic process
          nitric oxide biosynthetic process
          inflammatory response
          nitric oxide mediated signal transduction
          negative regulation of gene expression
          peptidyl-cysteine S-nitrosylation
          positive regulation of guanylate cyclase activity
          response to lipopolysaccharide
          cellular response to drug
          regulation of cell proliferation
          negative regulation of protein catabolic process
          defense response to bacterium
          negative regulation of blood pressure
          regulation of protein kinase activity
          positive regulation of vasodilation
          regulation of insulin secretion
          positive regulation of killing of cells of other organism
          oxidation-reduction process
          cellular response to lipopolysaccharide
          cellular response to interferon-gamma
          actin binding
          NADPH-hemoprotein reductase activity
          nitric-oxide synthase activity
          receptor binding
          iron ion binding
          protein binding
          calmodulin binding
          beta-catenin binding
          cAMP-dependent protein kinase regulator activity
          FMN binding
          oxidoreductase activity
          protein kinase binding
          heme binding
          tetrahydrobiopterin binding
          arginine binding
          protein homodimerization activity
          cadherin binding
          metal ion binding
          flavin adenine dinucleotide binding
          nitric-oxide synthase binding
          Hsp90 protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100010918.mm.1 NM_010927.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010918.mm.1 TC1100001171.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932117 - -

          Back To Top
           

          Citations (37)

          TLR9 signaling is essential for the innate NK cell response in murine cutaneous leishmaniasis.
          Authors
          Liese J, Schleicher U, Bogdan C
          Journal
          Eur J Immunol - Page No 3424 (Sat Dec 01 00:00:00 EST 2007)
          Volume (Year)
          37 (2007)
          Product Usage
          -
          Loss of Nrf2 markedly exacerbates nonalcoholic steatohepatitis.
          Authors
          Chowdhry S, Nazmy MH, Meakin PJ, Dinkova-Kostova AT, Walsh SV, Tsujita T, Dillon JF, Ashford ML, Hayes JD
          Journal
          Free Radic Biol Med - Page No 357 (Fri Jan 15 00:00:00 EST 2010)
          Volume (Year)
          48 (2010)
          Product Usage
          -
          Fucoxanthin regulates adipocytokine mRNA expression in white adipose tissue of diabetic/obese KK-Ay mice.
          Authors
          Hosokawa M, Miyashita T, Nishikawa S, Emi S, Tsukui T, Beppu F, Okada T, Miyashita K
          Journal
          Arch Biochem Biophys - Page No 17 (Wed Dec 01 00:00:00 EST 2010)
          Volume (Year)
          504 (2010)
          Product Usage
          -
          Cytokine-mediated control of lipopolysaccharide-induced activation of small intestinal epithelial cells.
          Authors
          Lotz M, Konig T, Menard S, Gutle D, Bogdan C, Hornef MW
          Journal
          Immunology - Page No 306 (Thu Nov 01 00:00:00 EDT 2007)
          Volume (Year)
          122 (2007)
          Product Usage
          -
          Immortalized mouse articular cartilage cell lines retain chondrocyte phenotype and respond to both anabolic factor BMP-2 and pro-inflammatory factor IL-1.
          Authors
          Majumdar MK, Chockalingam PS, Bhat RA, Sheldon R, Keohan C, Blanchet T, Glasson S, Morris EA
          Journal
          J Cell Physiol - Page No 68 (Tue Apr 01 00:00:00 EDT 2008)
          Volume (Year)
          215 (2008)
          Product Usage
          -
          Cytokines, signaling pathways, and effector molecules required for the control of Leishmania (Viannia) braziliensis in mice.
          Authors
          Rocha FJ, Schleicher U, Mattner J, Alber G, Bogdan C
          Journal
          Infect Immun - Page No 3823 (Wed Aug 01 00:00:00 EDT 2007)
          Volume (Year)
          75 (2007)
          Product Usage
          -
          Nitric oxide synthase modulates CFA-induced thermal hyperalgesia through cytokine regulation in mice.
          Authors
          Chen Y, Boettger MK, Reif A, Schmitt A, Uceyler N, Sommer C
          Journal
          Mol Pain - Page No 13 (Fri Jan 01 00:00:00 EST 2010)
          Volume (Year)
          6 (2010)
          Product Usage
          -
          High mobility group box 1 protein is released by neural cells upon different stresses and worsens ischemic neurodegeneration in vitro and in vivo.
          Authors
          Faraco G, Fossati S, Bianchi ME, Patrone M, Pedrazzi M, Sparatore B, Moroni F, Chiarugi A
          Journal
          J Neurochem - Page No 590 (Mon Oct 01 00:00:00 EDT 2007)
          Volume (Year)
          103 (2007)
          Product Usage
          -
          Congenital NOS2 deficiency prevents impairment of hypoxic pulmonary vasoconstriction in murine ventilator-induced lung injury.
          Authors
          Liu R, Hotta Y, Graveline AR, Evgenov OV, Buys ES, Bloch KD, Ichinose F, Zapol WM
          Journal
          Am J Physiol Lung Cell Mol Physiol - Page No L1300 (Thu Nov 01 00:00:00 EDT 2007)
          Volume (Year)
          293 (2007)
          Product Usage
          -
          Inducible nitric oxide synthase and estradiol exhibit complementary neuroprotective roles after ischemic brain injury.
          Authors
          Brown CM, Dela Cruz CD, Yang E, Wise PM
          Journal
          Exp Neurol - Page No 782 (Tue Apr 01 00:00:00 EDT 2008)
          Volume (Year)
          210 (2008)
          Product Usage
          -
          Central administration of lipopolysaccharide induces depressive-like behavior in vivo and activates brain indoleamine 2,3 dioxygenase in murine organotypic hippocampal slice cultures.
          Authors
          Fu X, Zunich SM, O'Connor JC, Kavelaars A, Dantzer R, Kelley KW
          Journal
          J Neuroinflammation - Page No 43 (Fri Jan 01 00:00:00 EST 2010)
          Volume (Year)
          7 (2010)
          Product Usage
          -
          Mutant alpha-synuclein overexpression mediates early proinflammatory activity.
          Authors
          Su X, Federoff HJ, Maguire-Zeiss KA
          Journal
          Neurotox Res - Page No 238 (Thu Oct 01 00:00:00 EDT 2009)
          Volume (Year)
          16 (2009)
          Product Usage
          -
          Low-Intensity Pulsed Ultrasound Modulates Shear Stress Induced PGHS-2 Expression and PGE(2) Synthesis in MLO-Y4 Osteocyte-Like Cells.
          Authors
          Saini V, Yadav S, McCormick S
          Journal
          Ann Biomed Eng - Page No (Sat Sep 04 00:00:00 EDT 2010)
          Volume (Year)
           (2010)
          Product Usage
          -
          Differential involvement of RelB in morphine-induced modulation of chemotaxis, NO, and cytokine production in murine macrophages and lymphocytes.
          Authors
          Martucci C, Franchi S, Lattuada D, Panerai AE, Sacerdote P
          Journal
          J Leukoc Biol - Page No 344 (Mon Jan 01 00:00:00 EST 2007)
          Volume (Year)
          81 (2007)
          Product Usage
          -
          Endothelial nitric oxide synthase deficiency in mice results in reduced chondrocyte proliferation and endochondral bone growth.
          Authors
          Yan Q, Feng Q, Beier F
          Journal
          Arthritis Rheum - Page No 2013 (Thu Jul 01 00:00:00 EDT 2010)
          Volume (Year)
          62 (2010)
          Product Usage
          -
          Adiponectin upregulates ferritin heavy chain in skeletal muscle cells.
          Authors
          Ikegami Y, Inukai K, Imai K, Sakamoto Y, Katagiri H, Kurihara S, Awata T, Katayama S
          Journal
          Diabetes - Page No 61 (Thu Jan 01 00:00:00 EST 2009)
          Volume (Year)
          58 (2009)
          Product Usage
          -
          Pioglitazone protects the myocardium against ischemia-reperfusion injury in eNOS and iNOS knockout mice.
          Authors
          Ye Y, Lin Y, Manickavasagam S, Perez-Polo JR, Tieu BC, Birnbaum Y
          Journal
          Am J Physiol Heart Circ Physiol - Page No H2436 (Mon Dec 01 00:00:00 EST 2008)
          Volume (Year)
          295 (2008)
          Product Usage
          -
          Suppressive effects of alloxanthin and diatoxanthin from Halocynthia roretzi on LPS-induced expression of pro-inflammatory genes in RAW264.7 cells.
          Authors
          Konishi I, Hosokawa M, Sashima T, Maoka T, Miyashita K
          Journal
          J Oleo Sci - Page No 181 (Tue Jan 01 00:00:00 EST 2008)
          Volume (Year)
          57 (2008)
          Product Usage
          -
          Endogenous interleukin-10 inhibits angiotensin II-induced vascular dysfunction.
          Authors
          Didion SP, Kinzenbaw DA, Schrader LI, Chu Y, Faraci FM
          Journal
          Hypertension - Page No 619 (Tue Sep 01 00:00:00 EDT 2009)
          Volume (Year)
          54 (2009)
          Product Usage
          -
          Psychological stress suppresses innate IFN-gamma production via glucocorticoid receptor activation: reversal by the anxiolytic chlordiazepoxide.
          Authors
          Curtin NM, Boyle NT, Mills KH, Connor TJ
          Journal
          Brain Behav Immun - Page No 535 (Fri May 01 00:00:00 EDT 2009)
          Volume (Year)
          23 (2009)
          Product Usage
          -
          Differential roles of nitric oxide synthase isozymes in cardiotoxicity and mortality following chronic doxorubicin treatment in mice.
          Authors
          Deng S, Kruger A, Schmidt A, Metzger A, Yan T, Godtel-Armbrust U, Hasenfuss G, Brunner F, Wojnowski L
          Journal
          Naunyn Schmiedebergs Arch Pharmacol - Page No 25 (Wed Jul 01 00:00:00 EDT 2009)
          Volume (Year)
          380 (2009)
          Product Usage
          -
          Modulation by budesonide of DNA methylation and mRNA expression in mouse lung tumors.
          Authors
          Pereira MA, Tao L, Liu Y, Li L, Steele VE, Lubet RA
          Journal
          Int J Cancer - Page No 1150 (Thu Mar 01 00:00:00 EST 2007)
          Volume (Year)
          120 (2007)
          Product Usage
          -
          Enhanced production of nitric oxide, reactive oxygen species, and pro-inflammatory cytokines in very long chain saturated fatty acid-accumulated macrophages.
          Authors
          Yanagisawa N, Shimada K, Miyazaki T, Kume A, Kitamura Y, Sumiyoshi K, Kiyanagi T, Iesaki T, Inoue N, Daida H
          Journal
          Lipids Health Dis - Page No 48 (Tue Jan 01 00:00:00 EST 2008)
          Volume (Year)
          7 (2008)
          Product Usage
          -
          Purple carrot (Daucus carota L.) polyacetylenes decrease lipopolysaccharide-induced expression of inflammatory proteins in macrophage and endothelial cells.
          Authors
          Metzger BT, Barnes DM, Reed JD
          Journal
          J Agric Food Chem - Page No 3554 (Wed May 28 00:00:00 EDT 2008)
          Volume (Year)
          56 (2008)
          Product Usage
          -
          Both TLR2 and TLR4 are required for the effective immune response in Staphylococcus aureus-induced experimental murine brain abscess.
          Authors
          Stenzel W, Soltek S, Sanchez-Ruiz M, Akira S, Miletic H, Schluter D, Deckert M
          Journal
          Am J Pathol - Page No 132 (Tue Jan 01 00:00:00 EST 2008)
          Volume (Year)
          172 (2008)
          Product Usage
          -
          The APOE4 genotype alters the response of microglia and macrophages to 17beta-estradiol.
          Authors
          Brown CM, Choi E, Xu Q, Vitek MP, Colton CA
          Journal
          Neurobiol Aging - Page No 1783 (Mon Dec 01 00:00:00 EST 2008)
          Volume (Year)
          29 (2008)
          Product Usage
          -
          The involvement of nitric oxide in the enhanced expression of mu-opioid receptors during intestinal inflammation in mice.
          Authors
          Pol O, Sasaki M, Jimenez N, Dawson VL, Dawson TM, Puig MM
          Journal
          Br J Pharmacol - Page No 758 (Fri Jul 01 00:00:00 EDT 2005)
          Volume (Year)
          145 (2005)
          Product Usage
          -
          Psychological stress reactivates dextran sulfate sodium-induced chronic colitis in mice.
          Authors
          Melgar S, Engstrom K, Jagervall A, Martinez V
          Journal
          Stress - Page No 348 (Tue Jan 01 00:00:00 EST 2008)
          Volume (Year)
          11 (2008)
          Product Usage
          -
          Arginine supplementation induces myoblast fusion via augmentation of nitric oxide production.
          Authors
          Long JH, Lira VA, Soltow QA, Betters JL, Sellman JE, Criswell DS
          Journal
          J Muscle Res Cell Motil - Page No 577 (Sun Jan 01 00:00:00 EST 2006)
          Volume (Year)
          27 (2006)
          Product Usage
          -
          Small interfering RNA (siRNA) delivery into murine bone marrow-derived macrophages by electroporation.
          Authors
          Wiese M, Castiglione K, Hensel M, Schleicher U, Bogdan C, Jantsch J
          Journal
          J Immunol Methods - Page No 102 (Sun Feb 28 00:00:00 EST 2010)
          Volume (Year)
          353 (2010)
          Product Usage
          -
          Regulation of the inflammatory response to Staphylococcus aureus-induced brain abscess by interleukin-10.
          Authors
          Stenzel W, Dahm J, Sanchez-Ruiz M, Miletic H, Hermann M, Courts C, Schwindt H, Utermohlen O, Schluter D, Deckert M
          Journal
          J Neuropathol Exp Neurol - Page No 1046 (Thu Dec 01 00:00:00 EST 2005)
          Volume (Year)
          64 (2005)
          Product Usage
          -
          Expression profiles of proliferative and antiapoptotic genes in sporadic and colitis-related mouse colon cancer models.
          Authors
          Svec J, Ergang P, Mandys V, Kment M, Pacha J
          Journal
          Int J Exp Pathol - Page No 44 (Mon Feb 01 00:00:00 EST 2010)
          Volume (Year)
          91 (2010)
          Product Usage
          -
          PKC412 (CGP41251) modulates the proliferation and lipopolysaccharide-induced inflammatory responses of RAW 264.7 macrophages.
          Authors
          Miyatake K, Inoue H, Hashimoto K, Takaku H, Takata Y, Nakano S, Yasui N, Itakura M
          Journal
          Biochem Biophys Res Commun - Page No 115 (Fri Aug 17 00:00:00 EDT 2007)
          Volume (Year)
          360 (2007)
          Product Usage
          -
          Intra-colonic administration of the TLR7 agonist R-848 induces an acute local and systemic inflammation in mice.
          Authors
          Karlsson A, Jagervall K, Utkovic H, Karlsson L, Rehnstrom E, Fredin MF, Gillberg PG, Jansson L, Michaelsson E, Melgar S
          Journal
          Biochem Biophys Res Commun - Page No 242 (Fri Mar 07 00:00:00 EST 2008)
          Volume (Year)
          367 (2008)
          Product Usage
          -
          Discrepancy between the in vitro and in vivo effects of murine mesenchymal stem cells on T-cell proliferation and collagen-induced arthritis.
          Authors
          Schurgers E, Kelchtermans H, Mitera T, Geboes L, Matthys P
          Journal
          Arthritis Res Ther - Page No R31 (Fri Jan 01 00:00:00 EST 2010)
          Volume (Year)
          12 (2010)
          Product Usage
          -
          IL-4(-/-) mice with lethal Mesocestoides corti infections--reduced Th2 cytokines and alternatively activated macrophages.
          Authors
          O'Connell AE, Kerepesi LA, Vandergrift GL, Herbert DR, VAN Winkle TJ, Hooper DC, Pearce EJ, Abraham D
          Journal
          Parasite Immunol - Page No 741 (Tue Dec 01 00:00:00 EST 2009)
          Volume (Year)
          31 (2009)
          Product Usage
          -
          Immortalized cell lines from mouse xiphisternum preserve chondrocyte phenotype.
          Authors
          Majumdar MK, Chockalingam PS, Bhat RA, Sheldon R, Keohan C, Blanchet T, Glasson S, Morris EA
          Journal
          J Cell Physiol - Page No 551 (Wed Nov 01 00:00:00 EST 2006)
          Volume (Year)
          209 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline