Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440546_s1
          See other NPY6R GE Assays ›
          Gene Symbol:
          Npy6r
          Gene Name:
          neuropeptide Y receptor Y6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 44270127 - 44277700 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440546_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Npy6r
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010935.3 NP_035065.1 2 1132 97
          GenBank mRNA AK030279.1 - 2 1134 97
          BC103620.1 - 2 990 40046295 97
          BC103621.1 - 2 990 40046291 97
          BC103622.1 - 2 990 40046297 97
          BC103667.1 - 2 990 40046294 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000453.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Npy6r

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          chemical synaptic transmission
          peptide YY receptor activity
          signal transducer activity
          G-protein coupled receptor activity
          neuropeptide Y receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113021 AK030279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2113021 NM_010935.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82036 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122001 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113021 AK030279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2113021 NM_010935.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291323 BC103622.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291323 BC103667.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291323 AK030279.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291323 BC103620.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P291323 BC103621.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665914 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08983867 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122001 - -
          Whole Mouse Genome G4122A A_51_P291323 BC103622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291323 BC103667.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291323 AK030279.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291323 BC103620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291323 BC103621.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139507 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13955756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14084623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13955747 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13955746 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13955759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14084621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31666004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31665982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14084620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14116800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13680916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13680907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13680911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13828781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14033217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13924873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14033219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665896 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455458 - -
          Affymetrix HT MG-430 PM Array Plate 1422347_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438086_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455457 - -
          Mouse Genome 430 2.0 Array 1438086_PM_at - -
          Mouse Genome 430 2.0 Array 1438086_at - -
          Mouse Genome 430 2.0 Array 1422347_PM_at - -
          Mouse Genome 430 2.0 Array 1422347_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18169_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18169_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800003332.mm.1 NM_010935.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800003332.mm.1 NM_010935.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800003332.mm.1 TC1800000453.mm.1 MTA TC
          Mouse Expression Set 430 1422347_PM_at - -
          Mouse Expression Set 430 1422347_at - -
          Mouse Expression Set 430 1438086_at - -
          Mouse Expression Set 430 1438086_PM_at - -
          Murine Genome U74v2 Set 99811_at - -
          Murine Genome U74v2 Set 99811_PM_at - -
          Murine Genome U74v2 Set 168298_at - -
          Murine Genome U74v2 Set 168298_PM_at - -
          Mouse Genome 430A 2.0 Array 1422347_PM_at - -
          Mouse Genome 430A 2.0 Array 1422347_at - -
          Affymetrix HT Mouse Genome 430A Array 1422347_at - -
          Affymetrix HT Mouse Genome 430A Array 1422347_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438086_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438086_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 831897 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4560136 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004560136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655419 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655419 - -
          mouse-6 v1.1 (Illumina) scl52010.2.1_247-S - -
          Illumina Sentrix mouse-6.v1 bead array 4560136 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52010.2.1_247-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4560136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2655419 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2655419 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline