Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440616_g1
          See other REG3B GE Assays ›
          Gene Symbol:
          Reg3b
          Gene Name:
          regenerating islet-derived 3 beta
          Gene Aliases:
          HIP, PAP1, Pap, REG-III
          Chromosome Location:
          Chr.6: 78370885 - 78373466 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440616_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Reg3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011036.1 NP_035166.1 5 - 6 492 73
          GenBank mRNA BC027525.1 - 4 - 5 475 3471932 73
          D13509.1 - 465 73
          D63359.1 - 5 - 6 492 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000875.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Reg3b

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          acute-phase response
          inflammatory response
          positive regulation of cell proliferation
          negative regulation of neuron apoptotic process
          defense response to Gram-negative bacterium
          defense response to Gram-positive bacterium
          negative regulation of cell death
          negative regulation of hydrogen peroxide-induced neuron death
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169671 NM_011036.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169671 BC027525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169671 D63359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169671 NM_011036.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169671 BC027525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P169671 D63359.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319454 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169671 NM_011036.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169671 BC027525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169671 D63359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169671 NM_011036.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169671 BC027525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P169671 D63359.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169671 NM_011036.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169671 BC027525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169671 D63359.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169671 NM_011036.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169671 BC027525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169671 D63359.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24569769 - -
          Whole Mouse Genome G4122A A_51_P169671 NM_011036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169671 BC027525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169671 D63359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169671 NM_011036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169671 BC027525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169671 D63359.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05423256 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05423245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05423274 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05423280 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05423278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24569769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05081884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24569784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10539185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10539180 - -
          Affymetrix HT MG-430 PM Array Plate 1416297_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1448290_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10539179 - -
          Mouse Genome 430 2.0 Array 1416297_s_at - -
          Mouse Genome 430 2.0 Array 1416297_PM_s_at - -
          Mouse Genome 430 2.0 Array 1448290_at - -
          Mouse Genome 430 2.0 Array 1448290_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18489_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18489_at - -
          Affymetrix Mouse Clariom S Array PSR0600006665.mm.1 TC0600000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600006665.mm.1 NM_011036.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006665.mm.1 NM_011036.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006666.mm.1 TC0600000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600006666.mm.1 NM_011036.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600006666.mm.1 NM_011036.1 Refseq
          Mouse Expression Set 430 1448290_PM_at - -
          Mouse Expression Set 430 1416297_PM_s_at - -
          Mouse Expression Set 430 1416297_s_at - -
          Mouse Expression Set 430 1448290_at - -
          Murine Genome U74v2 Set 161890_PM_f_at - -
          Murine Genome U74v2 Set 161890_f_at - -
          Murine Genome U74v2 Set 96009_PM_s_at - -
          Murine Genome U74v2 Set 96009_s_at - -
          Mouse Genome 430A 2.0 Array 1448290_at - -
          Mouse Genome 430A 2.0 Array 1416297_s_at - -
          Mouse Genome 430A 2.0 Array 1448290_PM_at - -
          Mouse Genome 430A 2.0 Array 1416297_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1448290_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416297_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416297_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1448290_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 671539 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1780593 - -
          Illumina mouse-6 v1.0 expression beadchip 2690524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2630391 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2705804 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2705804 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2630391 - -
          mouse-6 v1.1 (Illumina) scl29870.6.1_39-S - -
          mouse-6 v1.1 (Illumina) scl0001020.1_68-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690524 - -
          Illumina Sentrix mouse-6.v1 bead array 1780593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001020.1_68-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29870.6.1_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690524 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2705804 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2887421 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2705804 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2887421 - -

          Back To Top
           

          Citations (1)

          Interleukin-18, together with interleukin-12, induces severe acute pancreatitis in obese but not in nonobese leptin-deficient mice.
          Authors
          Sennello JA, Fayad R, Pini M, Gove ME, Ponemone V, Cabay RJ, Siegmund B, Dinarello CA, Fantuzzi G
          Journal
          Proc Natl Acad Sci U S A - Page No 8085 (Tue Jun 10 00:00:00 EDT 2008)
          Volume (Year)
          105 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline