Search
Search

Scn1b| Gene Symbol: Scn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011322.2 | NP_035452.1 | 3 - 4 | 690 | 86 | |
| GenBank mRNA | BC009652.1 | - | 3 - 4 | 690 | 3489368 | 86 |
| BC039140.1 | - | 4 - 5 | 693 | 3672893 | 86 | |
| U46681.1 | - | 3 - 4 | 468 | 86 | ||
| U85786.1 | - | 3 - 4 | 510 | 86 | ||
Set Membership: |
|