Search
Search

Sema6c| Gene Symbol: Sema6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272024.1 | NP_001258953.1 | 9 - 10 | 1042 | 55 | |
| NM_011351.2 | NP_035481.1 | 6 - 7 | 610 | 55 | ||
| NR_073549.1 | 7 - 8 | 935 | 55 | |||
| NR_073550.1 | 7 - 8 | 949 | 55 | |||
| NR_073551.1 | 8 - 9 | 998 | 55 | |||
| GenBank mRNA | AB013729.1 | - | 6 - 7 | 575 | 55 | |
| AF363973.1 | - | 6 - 7 | 643 | 55 | ||
| AK031131.1 | - | 7 - 8 | 847 | 55 | ||
| AK129459.1 | - | 1 - 2 | 86 | 55 | ||
| AK161042.1 | - | 1 - 2 | 89 | 55 | ||
| AK165085.1 | - | 7 - 8 | 862 | 55 | ||
| BC051979.1 | - | 8 - 9 | 904 | 55 | ||
| BC059007.1 | - | 9 - 10 | 957 | 55 | ||
Set Membership: |
|