Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441589_m1
          See other STXBP2 GE Assays ›
          Gene Symbol:
          Stxbp2
          Gene Name:
          syntaxin binding protein 2
          Gene Aliases:
          C79054, Munc-18-2, Munc-18b, Munc18b, Sxtbp2, Sxtp2, Unc18-2, Unc18b, muSec1
          Chromosome Location:
          Chr.8: 3630960 - 3643707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441589_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stxbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011503.4 NP_035633.1 14 - 15 1252 63
          GenBank mRNA AF263345.1 - 14 - 15 1259 63
          AK088364.1 - 14 - 15 1274 63
          AK133419.1 - 14 - 15 1253 63
          BC003477.1 - 14 - 15 1242 63
          BC008102.1 - 14 - 15 1262 3966319 63
          D42068.1 - 14 - 15 1244 63
          U19520.1 - 14 - 15 1266 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000017.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stxbp2

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte mediated cytotoxicity
          transport
          exocytosis
          vesicle docking involved in exocytosis
          protein transport
          vesicle-mediated transport
          regulation of mast cell degranulation
          syntaxin-1 binding
          syntaxin binding
          syntaxin-3 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614885 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614884 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134062 BC008102.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134062 D42068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134062 U19520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134062 AK133419.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P134062 BC003477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000668 AK133419.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2000668 NM_011503.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539560 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174191 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000668 AK133419.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2000668 NM_011503.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P134062 BC008102.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P134062 D42068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P134062 U19520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P134062 AK133419.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P134062 BC003477.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614887 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03810478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03810489 - -
          Whole Mouse Genome G4122A A_51_P134070 BC008102.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134070 D42068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134070 U19520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134070 AK133419.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134070 AK184114.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127008 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115048 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03810478 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614933 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569849 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569851 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569850 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569856 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569857 - -
          Affymetrix HT MG-430 PM Array Plate 1448341_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1416449_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1437048_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569848 - -
          Mouse Genome 430 2.0 Array 1437048_PM_x_at - -
          Mouse Genome 430 2.0 Array 1448341_a_at - -
          Mouse Genome 430 2.0 Array 1416449_PM_x_at - -
          Mouse Genome 430 2.0 Array 1416449_x_at - -
          Mouse Genome 430 2.0 Array 1437048_x_at - -
          Mouse Genome 430 2.0 Array 1448341_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20911_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20911_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa15700_at - -
          Affymetrix Mouse Clariom S Array PSR0800000246.mm.1 NM_011503.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000246.mm.1 NM_011503.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000246.mm.1 TC0800000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000245.mm.1 NM_011503.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000245.mm.1 NM_011503.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000245.mm.1 TC0800000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000247.mm.1 NM_011503.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000247.mm.1 NM_011503.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000247.mm.1 TC0800000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000253.mm.1 NM_011503.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000253.mm.1 NM_011503.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000253.mm.1 TC0800000017.mm.1 MTA TC
          Mouse Expression Set 430 1448341_PM_a_at - -
          Mouse Expression Set 430 1437048_PM_x_at - -
          Mouse Expression Set 430 1437048_x_at - -
          Mouse Expression Set 430 1416449_x_at - -
          Mouse Expression Set 430 1416449_PM_x_at - -
          Mouse Expression Set 430 1448341_a_at - -
          Murine Genome U74v2 Set 101398_PM_at - -
          Murine Genome U74v2 Set 162272_r_at - -
          Murine Genome U74v2 Set 101398_at - -
          Murine Genome U74v2 Set 162272_PM_r_at - -
          Murine Genome U74v2 Set 169529_PM_i_at - -
          Murine Genome U74v2 Set 169529_i_at - -
          Mouse Genome 430A 2.0 Array 1437048_x_at - -
          Mouse Genome 430A 2.0 Array 1448341_a_at - -
          Mouse Genome 430A 2.0 Array 1416449_x_at - -
          Mouse Genome 430A 2.0 Array 1448341_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416449_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437048_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448341_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448341_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437048_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416449_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437048_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416449_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 365065 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2360594 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240462 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240462 - -
          mouse-6 v1.1 (Illumina) scl34132.19.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360594 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34132.19.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2657728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2657728 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34132.19.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline