Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441638_m1
          See other ABCC9 GE Assays ›
          Gene Symbol:
          Abcc9
          Gene Name:
          ATP-binding cassette, sub-family C (CFTR/MRP), member 9
          Gene Aliases:
          AI414027, AI449286, SUR2A, SUR2B, Sur2
          Chromosome Location:
          Chr.6: 142587862 - 142702760 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441638_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcc9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044720.1 NP_001038185.1 30 - 31 3841 73
          NM_011511.2 NP_035641.1 31 - 32 3880 73
          NM_021041.2 NP_066378.1 31 - 32 3880 73
          NM_021042.2 NP_066379.2 30 - 31 3775 73
          GenBank mRNA AF003531.1 - 30 - 31 3775 73
          AK052291.1 - 5 - 6 462 73
          D86037.2 - 31 - 32 3824 73
          D86038.2 - 31 - 32 3824 73
          U97066.1 - 31 - 32 3880 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003379.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Recommended Assay Mus musculus Mouse ABC Transporters 1700 Inventoried Mouse_ABC_Transporters Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcc9

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          potassium ion transport
          signal transduction
          potassium ion import
          defense response to virus
          transmembrane transport
          nucleotide binding
          potassium channel activity
          ATP binding
          drug binding
          sulfonylurea receptor activity
          potassium channel regulator activity
          ATPase activity
          syntaxin binding
          ATPase activity, coupled to transmembrane movement of substances
          identical protein binding
          ion channel binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 TC0600003379.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_001044720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_001044720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_011511.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_011511.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_021041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_021041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_021042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026630.mm.1 NM_021042.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 TC0600003379.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_001044720.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_001044720.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_011511.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_011511.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_021041.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_021041.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_021042.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600026631.mm.1 NM_021042.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 589278 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0