Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00442792_m1
          See other FCER2A GE Assays ›
          Gene Symbol:
          Fcer2a
          Gene Name:
          Fc receptor, IgE, low affinity II, alpha polypeptide
          Gene Aliases:
          CD23, Fce2, Fcer2, Ly-42
          Chromosome Location:
          Chr.8: 3680726 - 3694174 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00442792_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fcer2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253737.1 NP_001240666.1 8 - 9 568 68
          NM_001253739.1 NP_001240668.1 8 - 9 579 68
          NM_001253743.1 NP_001240672.1 9 - 10 628 68
          NM_001253745.1 NP_001240674.1 7 - 8 505 68
          NM_001253746.1 NP_001240675.1 7 - 8 505 68
          NM_001253747.1 NP_001240676.1 8 - 9 589 68
          NM_013517.4 NP_038545.1 9 - 10 709 68
          GenBank mRNA AK156654.1 - 9 - 10 712 68
          AK172591.1 - 9 - 10 712 68
          AY069980.1 - 7 - 8 471 68
          AY069981.1 - 7 - 8 471 68
          BC119164.1 - 8 - 9 576 8733913 68
          BC119190.1 - 8 - 9 576 8733939 68
          M34163.1 - 9 - 10 711 68
          M99371.1 - 9 - 10 701 68
          S67072.1 - 8 - 9 560 68
          S67073.1 - 7 - 8 440 68
          S67074.1 - 8 - 9 479 68
          X64223.1 - 8 - 9 568 68
          X64224.1 - 8 - 9 579 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001616.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fcer2a

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of humoral immune response mediated by circulating immunoglobulin
          positive regulation of nitric-oxide synthase activity
          positive regulation of killing of cells of other organism
          positive regulation of nitric-oxide synthase biosynthetic process
          IgE binding
          carbohydrate binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117345 M99371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117345 AK156654.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117345 AK172591.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2117345 NM_013517.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 63772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937780 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 457916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35231 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102097 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138282 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117345 M99371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117345 AK156654.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117345 AK172591.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117345 NM_013517.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P210591 M99371.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P210591 M34163.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P210591 NM_013517.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25615231 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01662869 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102097 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138282 - -
          Whole Mouse Genome G4122A A_51_P210591 M99371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210591 M34163.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P210591 NM_013517.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103671 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138282 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102097 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06929235 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06929228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25615253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07263984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07022513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06986160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06929228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07183924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07326370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06929232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07314756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07263989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06986162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06986151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07022511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07314758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25615180 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576771 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576766 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576764 - -
          Affymetrix HT MG-430 PM Array Plate 1422122_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451713_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576757 - -
          Mouse Genome 430 2.0 Array 1451713_a_at - -
          Mouse Genome 430 2.0 Array 1422122_at - -
          Mouse Genome 430 2.0 Array 1422122_PM_at - -
          Mouse Genome 430 2.0 Array 1451713_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14128_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14128_at - -
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253737.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253737.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253739.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253739.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253743.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253743.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253746.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253746.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253747.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_001253747.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_013517.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 NM_013517.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013283.mm.1 TC0800001616.mm.1 MTA TC
          Mouse Expression Set 430 1451713_a_at - -
          Mouse Expression Set 430 1451713_PM_a_at - -
          Mouse Expression Set 430 1422122_at - -
          Mouse Expression Set 430 1422122_PM_at - -
          Murine Genome U74v2 Set 94141_PM_at - -
          Murine Genome U74v2 Set 94141_at - -
          Mouse Genome 430A 2.0 Array 1451713_a_at - -
          Mouse Genome 430A 2.0 Array 1422122_PM_at - -
          Mouse Genome 430A 2.0 Array 1451713_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422122_at - -
          Affymetrix HT Mouse Genome 430A Array 1422122_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451713_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422122_at - -
          Affymetrix HT Mouse Genome 430A Array 1451713_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5720403 - -
          Illumina mouse-6 v1.0 expression beadchip 5550242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2652877 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246358 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246358 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2652877 - -
          mouse-6 v1.1 (Illumina) scl014128.1_25-S - -
          mouse-6 v1.1 (Illumina) scl0014128.2_101-S - -
          Illumina Sentrix mouse-6.v1 bead array 5720403 - -
          Illumina Sentrix mouse-6.v1 bead array 5550242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014128.1_25-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014128.2_101-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550242 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2975312 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2652875 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2652877 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2975312 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2652875 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2652877 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline