Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00443100_m1
          See other PVALB GE Assays ›
          Gene Symbol:
          Pvalb
          Gene Name:
          parvalbumin
          Gene Aliases:
          PV, Parv, Pva
          Chromosome Location:
          Chr.15: 78191117 - 78206351 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00443100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pvalb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013645.3 NP_038673.2 1 - 2 84 77
          GenBank mRNA AK002354.1 - 1 - 2 92 77
          AK005371.1 - 1 - 2 82 77
          AK013561.1 - 1 - 2 76 77
          AK131986.1 - 1 - 2 154 77
          BC027424.1 - 1 - 2 84 4925213 77
          S75909.1 - 2 - 3 145 77
          S75910.1 - 1021 77
          X54613.1 - 1 - 2 62 77
          X59382.1 - 2 - 3 118 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001827.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pvalb

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytosolic calcium ion concentration
          calcium ion binding
          protein homodimerization activity
          metal ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059040 BC027424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059040 NM_013645.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114437 X54613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114437 X59382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114437 AK131986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114437 BC027424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114437 X67141.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69337 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162361 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101511 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114437 X54613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114437 X59382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114437 AK131986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114437 BC027424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114437 X67141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059040 BC027424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059040 NM_013645.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P500156 BC027424.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P500156 X67141.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P500156 NM_013645.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P500156 AK013561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500156 BC027424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500156 X67141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500156 NM_013645.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P500156 AK013561.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124037 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12305857 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12305872 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12305888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12305860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12305908 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430298 - -
          Affymetrix HT MG-430 PM Array Plate 1417653_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430297 - -
          Mouse Genome 430 2.0 Array 1417653_PM_at - -
          Mouse Genome 430 2.0 Array 1417653_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19293_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19293_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500014270.mm.1 NM_013645.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014270.mm.1 TC1500001827.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500014269.mm.1 NM_013645.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500014269.mm.1 TC1500001827.mm.1 MTA TC
          Mouse Expression Set 430 1417653_at - -
          Mouse Expression Set 430 1417653_PM_at - -
          Murine Genome U74v2 Set 96720_f_at - -
          Murine Genome U74v2 Set 96719_i_at - -
          Murine Genome U74v2 Set 96719_PM_i_at - -
          Murine Genome U74v2 Set 96720_PM_f_at - -
          Murine Genome U74v2 Set 168316_i_at - -
          Murine Genome U74v2 Set 168316_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1417653_PM_at - -
          Mouse Genome 430A 2.0 Array 1417653_at - -
          Affymetrix HT Mouse Genome 430A Array 1417653_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417653_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 407684 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218223 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218223 - -
          mouse-6 v1.1 (Illumina) scl46996.5.2_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46996.5.2_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218223 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218223 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46996.5.2_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline