Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444381_m1
          See other PBSN GE Assays ›
          Gene Symbol:
          Pbsn
          Gene Name:
          probasin
          Gene Aliases:
          PB, Prbs
          Chromosome Location:
          Chr.X: 77837898 - 77853500 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444381_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pbsn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017471.2 NP_059499.1 5 - 6 509 80
          GenBank mRNA AF005204.1 - 5 - 6 513 80
          AK137501.1 - 5 - 6 536 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002507.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pbsn

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          transporter activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085365 NM_017471.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085365 AF005204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085365 AK137501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085368 NM_017471.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085368 AF005204.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085368 AK137501.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 659749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54498 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101669 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117180 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085368 NM_017471.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085368 AF005204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085368 AK137501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085365 NM_017471.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085365 AF005204.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085365 AK137501.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273790 NM_017471.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273790 AF005204.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273790 AK137501.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32479746 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09337290 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117180 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03701552 - -
          Whole Mouse Genome G4122A A_51_P273790 NM_017471.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273790 AF005204.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273790 AK137501.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130025 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117180 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14741062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14741064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14741056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14741060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32479663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14760656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14741069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14887387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32479747 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605478 - -
          Affymetrix HT MG-430 PM Array Plate 1449320_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605475 - -
          Mouse Genome 430 2.0 Array 1449320_at - -
          Mouse Genome 430 2.0 Array 1449320_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54192_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54192_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00016001.mm.1 NM_017471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016001.mm.1 NM_017471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016001.mm.1 TC0X00002507.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016005.mm.1 NM_017471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016005.mm.1 NM_017471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016005.mm.1 TC0X00002507.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016002.mm.1 NM_017471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016002.mm.1 NM_017471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016002.mm.1 TC0X00002507.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016006.mm.1 NM_017471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016006.mm.1 NM_017471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016006.mm.1 TC0X00002507.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016004.mm.1 NM_017471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016004.mm.1 NM_017471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016004.mm.1 TC0X00002507.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016003.mm.1 NM_017471.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016003.mm.1 NM_017471.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016003.mm.1 TC0X00002507.mm.1 MTA TC
          Mouse Expression Set 430 1449320_PM_at - -
          Mouse Expression Set 430 1449320_at - -
          Murine Genome U74v2 Set 94632_at - -
          Murine Genome U74v2 Set 94632_PM_at - -
          Mouse Genome 430A 2.0 Array 1449320_PM_at - -
          Mouse Genome 430A 2.0 Array 1449320_at - -
          Affymetrix HT Mouse Genome 430A Array 1449320_at - -
          Affymetrix HT Mouse Genome 430A Array 1449320_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 775949 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6020546 - -
          Illumina mouse-6 v1.0 expression beadchip 6020546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692759 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692759 - -
          mouse-6 v1.1 (Illumina) scl54104.7.7_144-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020546 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54104.7.7_144-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692759 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692759 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline