Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444450_m1
          See other APBA3 GE Assays ›
          Gene Symbol:
          Apba3
          Gene Name:
          amyloid beta (A4) precursor protein-binding, family A, member 3
          Gene Aliases:
          AW208791, Mint-3, X11gamma, lin-10, mint3
          Chromosome Location:
          Chr.10: 81266965 - 81273247 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444450_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apba3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018758.2 NP_061228.1 3 - 4 877 126
          GenBank mRNA AF070975.1 - 3 - 4 831 126
          AK045965.1 - 3 - 4 878 126
          AK082046.1 - 797 126
          AK170182.1 - 3 - 4 825 126
          BC005423.1 - 3 - 4 809 3487448 126
          BC009666.1 - 3 - 4 821 3490710 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000891.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apba3

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          transport
          chemical synaptic transmission
          regulation of gene expression
          protein transport
          negative regulation of catalytic activity
          beta-amyloid binding
          enzyme inhibitor activity
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980981 BC009666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980981 BQ031518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980981 AK169462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980981 AK170182.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980981 BC005423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977538 BC009666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977538 BQ031518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977538 AK169462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977538 AK170182.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977538 BC005423.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 506897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866057 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134012 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980981 BC009666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980981 BQ031518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980981 AK169462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980981 AK170182.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980981 BC005423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977538 BC009666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977538 BQ031518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977538 AK169462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977538 AK170182.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977538 BC005423.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134012 - -
          Whole Mouse Genome G4122A A_51_P285960 BC009666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285960 BQ031518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285960 AK169462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285960 AK170182.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285960 BC005423.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08624232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08624243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08624235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08624242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08624238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288501 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10365061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365065 - -
          Affymetrix HT MG-430 PM Array Plate 1416182_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10365056 - -
          Mouse Genome 430 2.0 Array 1454720_PM_at - -
          Mouse Genome 430 2.0 Array 1416182_PM_at - -
          Mouse Genome 430 2.0 Array 1454720_at - -
          Mouse Genome 430 2.0 Array 1416182_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57267_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57267_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000006977.mm.1 NM_018758.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006977.mm.1 NM_018758.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006977.mm.1 TC1000000891.mm.1 MTA TC
          Mouse Expression Set 430 1416182_at - -
          Mouse Expression Set 430 1416182_PM_at - -
          Mouse Expression Set 430 1454720_at - -
          Mouse Expression Set 430 1454720_PM_at - -
          Murine Genome U74v2 Set 102750_PM_at - -
          Murine Genome U74v2 Set 102750_at - -
          Mouse Genome 430A 2.0 Array 1416182_at - -
          Mouse Genome 430A 2.0 Array 1416182_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416182_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416182_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 536099 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2510348 - -
          Illumina mouse-6 v1.0 expression beadchip 2510348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002510348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2660110 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2660110 - -
          mouse-6 v1.1 (Illumina) scl057267.10_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 2510348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl057267.10_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2510348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2660110 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2660110 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl057267.10_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline