Search
Search

Fxyd1| Gene Symbol: Fxyd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019503.4 | NP_062376.2 | 5 - 6 | 310 | 72 | |
| NM_052991.4 | NP_443717.1 | 5 - 6 | 293 | 72 | ||
| NM_052992.3 | NP_443718.1 | 5 - 6 | 374 | 72 | ||
| NM_194321.2 | NP_919302.1 | 5 - 6 | 357 | 72 | ||
| GenBank mRNA | AA466595.1 | - | 5 - 6 | 310 | 72 | |
| AF089734.1 | - | 5 - 6 | 236 | 72 | ||
| AI853335.1 | - | 3 - 4 | 72 | |||
| AK002585.1 | - | 5 - 6 | 297 | 72 | ||
| AK003504.1 | - | 1 - 2 | 92 | 72 | ||
| BC024671.1 | - | 5 - 6 | 247 | 4208962 | 72 | |
| BY064152.1 | - | 5 - 6 | 309 | 72 | ||
| BY132051.1 | - | 5 - 6 | 279 | 72 | ||
| CA459152.1 | - | 5 - 6 | 315 | 72 | ||
| CK793316.1 | - | 5 - 6 | 292 | 72 | ||
| W16239.1 | - | 5 - 6 | 273 | 72 | ||
| W98524.1 | - | 5 - 6 | 288 | 71 | ||
Set Membership: |
|