Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444822_g1
          See other NME3 GE Assays ›
          Gene Symbol:
          Nme3
          Gene Name:
          NME/NM23 nucleoside diphosphate kinase 3
          Gene Aliases:
          1810009F08Rik, AI413736, DR-nm23, Ndk3, Nm23-DR, Nm23-M3
          Chromosome Location:
          Chr.17: 24896500 - 24897531 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444822_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nme3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019730.2 NP_062704.2 4 - 5 400 73
          GenBank mRNA AF153449.1 - 4 - 5 412 73
          AF288689.1 - 4 - 5 73
          AK007399.1 - 4 - 5 400 73
          BC028503.1 - 4 - 5 391 3326648 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002771.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nme3

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleotide metabolic process
          nucleoside diphosphate phosphorylation
          GTP biosynthetic process
          pyrimidine nucleotide metabolic process
          UTP biosynthetic process
          CTP biosynthetic process
          nucleotide metabolic process
          nucleoside triphosphate biosynthetic process
          phosphorylation
          regulation of apoptotic process
          nucleotide binding
          nucleoside diphosphate kinase activity
          ATP binding
          kinase activity
          transferase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100170 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100172 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100174 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100171 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100173 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 NM_019730.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AF153449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AF288689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AK002525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AK007399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 NM_019730.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AF153449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AF288689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AK002525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P395111 AK007399.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152057 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 NM_019730.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AF153449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AF288689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AK002525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AK007399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 NM_019730.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AF153449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AF288689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AK002525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P395111 AK007399.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 NM_019730.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AF153449.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AF288689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AK002525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AK007399.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 NM_019730.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AF153449.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AF288689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AK002525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P395111 AK007399.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100183 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152057 - -
          Whole Mouse Genome G4122A A_51_P395111 NM_019730.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AF153449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AF288689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AK002525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AK007399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 NM_019730.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AF153449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AF288689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AK002525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395111 AK007399.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140566 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230925 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100172 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100174 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100171 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100173 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13511856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442644 - -
          Affymetrix HT MG-430 PM Array Plate 1448905_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442643 - -
          Mouse Genome 430 2.0 Array 1460628_at - -
          Mouse Genome 430 2.0 Array 1460628_PM_at - -
          Mouse Genome 430 2.0 Array 1448905_PM_at - -
          Mouse Genome 430 2.0 Array 1448905_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79059_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79059_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700002860.mm.1 NM_019730.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002860.mm.1 NM_019730.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002860.mm.1 TC1700002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002857.mm.1 NM_019730.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002857.mm.1 NM_019730.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002857.mm.1 TC1700002771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002858.mm.1 NM_019730.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002858.mm.1 NM_019730.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002858.mm.1 TC1700002771.mm.1 MTA TC
          Mouse Expression Set 430 1448905_at - -
          Mouse Expression Set 430 1448905_PM_at - -
          Murine Genome U74v2 Set 94981_PM_i_at - -
          Murine Genome U74v2 Set 94982_PM_f_at - -
          Murine Genome U74v2 Set 94982_f_at - -
          Murine Genome U74v2 Set 94981_i_at - -
          Mouse Genome 430A 2.0 Array 1448905_PM_at - -
          Mouse Genome 430A 2.0 Array 1448905_at - -
          Affymetrix HT Mouse Genome 430A Array 1448905_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448905_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 920635 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1660440 - -
          Illumina mouse-6 v1.0 expression beadchip 1660440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744414 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744414 - -
          mouse-6 v1.1 (Illumina) scl50997.3.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50997.3.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2744414 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2744414 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50997.3.1_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline