Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445109_m1
          See other RETNLA GE Assays ›
          Gene Symbol:
          Retnla
          Gene Name:
          resistin like alpha
          Gene Aliases:
          1810019L16Rik, Fizz-1, Fizz1, HIMF, RELM-alpha, RELMa, RELMalpha, Xcp2
          Chromosome Location:
          Chr.16: 48842552 - 48844461 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445109_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Retnla
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020509.3 NP_065255.2 2 - 3 229 86
          GenBank mRNA AF205951.1 - 1 - 2 162 86
          AF290872.1 - 2 - 3 267 86
          AF316397.2 - 2 - 3 171 86
          AF323082.1 - 2 - 3 239 86
          AK007553.1 - 2 - 3 174 86
          BC029248.1 - 1 - 2 169 4189285 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000698.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Retnla

          Gene Ontology Categories:

          Process(es)

          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 AK007553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 BC029248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 NM_020509.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 AF205951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 AF290872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 AK007553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 BC029248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 NM_020509.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 AF205951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P257951 AF290872.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133935 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133403 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 AK007553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 BC029248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 NM_020509.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 AF205951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 AF290872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 AK007553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 BC029248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 NM_020509.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 AF205951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P257951 AF290872.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30742759 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133403 - -
          Whole Mouse Genome G4122A A_52_P254817 AK007553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P254817 BC029248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P254817 NM_020509.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P254817 AF205951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P254817 AF290872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 AK007553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 BC029248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 NM_020509.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 AF205951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 AF290872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 AK007553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 BC029248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 NM_020509.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 AF205951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257951 AF290872.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101923 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133403 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12756046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12756045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12756043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30742795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12756046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12756037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12918724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12918727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13019687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12756043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30742742 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10436096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436097 - -
          Affymetrix HT MG-430 PM Array Plate 1449015_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10436095 - -
          Mouse Genome 430 2.0 Array 1449015_PM_at - -
          Mouse Genome 430 2.0 Array 1449015_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57262_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57262_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600006088.mm.1 NM_020509.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006088.mm.1 NM_020509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006088.mm.1 TC1600000698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006085.mm.1 NM_020509.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006085.mm.1 NM_020509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006085.mm.1 TC1600000698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600006087.mm.1 NM_020509.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006087.mm.1 NM_020509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600006087.mm.1 TC1600000698.mm.1 MTA TC
          Mouse Expression Set 430 1449015_at - -
          Mouse Expression Set 430 1449015_PM_at - -
          Mouse Genome 430A 2.0 Array 1449015_at - -
          Mouse Genome 430A 2.0 Array 1449015_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449015_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449015_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 925799 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3840138 - -
          Illumina mouse-6 v1.0 expression beadchip 3840138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840138 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226472 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226472 - -
          mouse-6 v1.1 (Illumina) scl057262.3_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl057262.3_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226472 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226472 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline