Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445555_m1
          See other TRAPPC4 GE Assays ›
          Gene Symbol:
          Trappc4
          Gene Name:
          trafficking protein particle complex 4
          Gene Aliases:
          1500017G03Rik, AI303617, HSPC172, PTD009, Sbd, Sbdn, Sdcbp2, TRS23
          Chromosome Location:
          Chr.9: 44403759 - 44407563 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445555_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trappc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021789.2 NP_068561.1 3 - 4 701 59
          GenBank mRNA AF233340.1 - 3 - 4 658 59
          AK005276.1 - 3 - 4 486 59
          AK032168.1 - 3 - 4 467 59
          BC038898.1 - 3 - 4 701 5401311 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002164.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trappc4

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ER to Golgi vesicle-mediated transport
          vesicle-mediated transport
          dendrite development
          positive regulation of GTPase activity
          neurotransmitter receptor biosynthetic process
          protein binding
          Rab guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467465 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467464 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467467 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 AF233340.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 AK005276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 AK032168.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 BC038898.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 NM_021789.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 AF233340.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 AK005276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 AK032168.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 BC038898.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407904 NM_021789.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719052 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635105 AJ000262.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635105 AJ002272.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635105 AK138436.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635105 AK196068.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635105 BC034089.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635105 BC053043.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635105 NM_010404.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35512 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132696 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 AF233340.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 AK005276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 AK032168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 BC038898.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 NM_021789.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 AF233340.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 AK005276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 AK032168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 BC038898.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407904 NM_021789.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26467453 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132696 - -
          Whole Mouse Genome G4122A A_51_P407904 AF233340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 AK005276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 AK032168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 BC038898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 NM_021789.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 AF233340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 AK005276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 AK032168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 BC038898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407904 NM_021789.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641172 AF233340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641172 AK005276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641172 AK032168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641172 BC038898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641172 NM_021789.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111181 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08093111 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07518233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08093114 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467465 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467464 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26467467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26467426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08061282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07710227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07710237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07710221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07639134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26467451 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592851 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592855 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592854 - -
          Affymetrix HT MG-430 PM Array Plate 1415674_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1429632_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10592850 - -
          Mouse Genome 430 2.0 Array 1415674_PM_a_at - -
          Mouse Genome 430 2.0 Array 1429632_at - -
          Mouse Genome 430 2.0 Array 1429632_PM_at - -
          Mouse Genome 430 2.0 Array 1415674_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60409_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60409_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900017978.mm.1 NM_021789.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017978.mm.1 NM_021789.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017978.mm.1 TC0900002164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017979.mm.1 NM_021789.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017979.mm.1 NM_021789.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017979.mm.1 TC0900002164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017980.mm.1 NM_021789.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017980.mm.1 NM_021789.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017980.mm.1 TC0900002164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017977.mm.1 NM_021789.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017977.mm.1 NM_021789.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017977.mm.1 TC0900002164.mm.1 MTA TC
          Mouse Expression Set 430 1415674_PM_a_at - -
          Mouse Expression Set 430 1415674_a_at - -
          Mouse Expression Set 430 1415674_PM_a_at - -
          Mouse Expression Set 430 1415674_a_at - -
          Murine Genome U74v2 Set 160361_PM_at - -
          Murine Genome U74v2 Set 160361_at - -
          Mouse Genome 430A 2.0 Array 1415674_a_at - -
          Mouse Genome 430A 2.0 Array 1415674_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415674_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429632_at - -
          Affymetrix HT Mouse Genome 430A Array 1415674_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429632_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1415674_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1415674_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 785123 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 520044 - -
          Illumina mouse-6 v1.0 expression beadchip 520044 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520044 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225507 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225507 - -
          mouse-6 v1.1 (Illumina) scl35956.2.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 520044 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35956.2.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520044 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225507 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2790573 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225507 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2790573 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35956.2.1_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline