Search
Search
| Gene Symbol: Retn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204959.1 | NP_001191888.1 | 2 - 3 | 218 | 80 | |
| NM_022984.4 | NP_075360.1 | 2 - 3 | 218 | 80 | ||
| GenBank mRNA | AF290870.1 | - | 2 - 3 | 211 | 80 | |
| AF323080.1 | - | 2 - 3 | 217 | 80 | ||
| AK131613.1 | - | 2 - 3 | 219 | 80 | ||
| BC051196.1 | - | 2 - 3 | 204 | 1448672 | 80 | |
Set Membership: |
|