Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00446387_m1
          See other COX4I2 GE Assays ›
          Gene Symbol:
          Cox4i2
          Gene Name:
          cytochrome c oxidase subunit IV isoform 2
          Gene Aliases:
          Cox4b, CoxIV-2
          Chromosome Location:
          Chr.2: 152754173 - 152765037 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00446387_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox4i2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053091.2 NP_444321.1 2 - 3 141 79
          GenBank mRNA AF271382.1 - 2 - 3 141 79
          BC049623.1 - 1 - 2 150 6532529 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002300.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox4i2

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          hydrogen ion transmembrane transport
          cytochrome-c oxidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054663 NM_053091.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054663 AF271382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2054663 BC049623.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192560 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64259 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171481 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123307 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054663 NM_053091.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054663 AF271382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2054663 BC049623.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P202942 NM_053091.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P202942 AF271382.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21751000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21750984 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171481 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04742668 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123307 - -
          Whole Mouse Genome G4122A A_51_P202942 NM_053091.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P202942 AF271382.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119395 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128625 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171481 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123307 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01676357 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02198704 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01676350 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01676358 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01231662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21751004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21750984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01552126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01948528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01960286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01552123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01676357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01375083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01676354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01176028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01629824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01231662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01676358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02212722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21750984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21751039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01422103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01636045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02262401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01406064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10477186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10477184 - -
          Affymetrix HT MG-430 PM Array Plate 1421373_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10477179 - -
          Mouse Genome 430 2.0 Array 1421373_PM_at - -
          Mouse Genome 430 2.0 Array 1421373_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 84682_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 84682_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200017978.mm.1 NM_053091.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017978.mm.1 NM_053091.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017978.mm.1 TC0200002300.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017980.mm.1 NM_053091.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017980.mm.1 NM_053091.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017980.mm.1 TC0200002300.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017981.mm.1 NM_053091.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017981.mm.1 NM_053091.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200017981.mm.1 TC0200002300.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200017979.mm.1 TC0200002300.mm.1 MTA TC
          Mouse Expression Set 430 1421373_PM_at - -
          Mouse Expression Set 430 1421373_at - -
          Mouse Genome 430A 2.0 Array 1421373_PM_at - -
          Mouse Genome 430A 2.0 Array 1421373_at - -
          Affymetrix HT Mouse Genome 430A Array 1421373_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421373_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 551351 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6370519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2612178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2612178 - -
          mouse-6 v1.1 (Illumina) scl20106.5.1_101-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20106.5.1_101-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2612178 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2612178 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl20106.5.1_101-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline