Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00446681_m1
          See other PDK2 GE Assays ›
          Gene Symbol:
          Pdk2
          Gene Name:
          pyruvate dehydrogenase kinase, isoenzyme 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 95026258 - 95041371 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00446681_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133667.2 NP_598428.2 4 - 5 636 72
          GenBank mRNA AF267660.1 - 4 - 5 597 72
          AK002647.1 - 4 - 5 580 72
          BC021764.1 - 4 - 5 590 3488813 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003583.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdk2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glucose metabolic process
          regulation of gluconeogenesis
          protein phosphorylation
          regulation of pH
          insulin receptor signaling pathway
          regulation of acetyl-CoA biosynthetic process from pyruvate
          regulation of cellular ketone metabolic process
          regulation of glucose metabolic process
          phosphorylation
          peptidyl-serine phosphorylation
          cellular response to nutrient
          cellular response to reactive oxygen species
          glucose homeostasis
          intrinsic apoptotic signaling pathway by p53 class mediator
          nucleotide binding
          protein serine/threonine kinase activity
          pyruvate dehydrogenase (acetyl-transferring) kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          protein complex binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036286 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426276 AF267660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426276 AF267660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011802 AF267660.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151370 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139517 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426276 AF267660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426276 AF267660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011802 AF267660.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426276 AF267660.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426276 AF267660.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036299 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07449391 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139517 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02400135 - -
          Whole Mouse Genome G4122A A_51_P426276 AF267660.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P426276 AF267660.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126630 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111183 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139517 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383667 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383683 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383677 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08978324 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09349542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08979463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09349547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09179616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09383673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09383683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09094766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09383685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09383665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09380289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08978327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09383677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08979465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09349541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28036274 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10390108 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390105 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390109 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390107 - -
          Affymetrix Mouse Gene 1.0 ST Array 10390115 - -
          Affymetrix HT MG-430 PM Array Plate 1448825_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10390103 - -
          Mouse Genome 430 2.0 Array 1448825_at - -
          Mouse Genome 430 2.0 Array 1448825_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18604_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18604_at - -
          Affymetrix Mouse Clariom S Array PSR1100032843.mm.1 NM_133667.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032843.mm.1 NM_133667.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032843.mm.1 TC1100003583.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032844.mm.1 NM_133667.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032844.mm.1 NM_133667.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032844.mm.1 TC1100003583.mm.1 MTA TC
          Mouse Expression Set 430 1448825_at - -
          Mouse Expression Set 430 1448825_PM_at - -
          Mouse Genome 430A 2.0 Array 1448825_at - -
          Mouse Genome 430A 2.0 Array 1448825_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448825_at - -
          Affymetrix HT Mouse Genome 430A Array 1448825_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 917445 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2690017 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690017 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737658 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737658 - -
          mouse-6 v1.1 (Illumina) scl39696.12_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690017 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39696.12_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690017 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2737658 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737658 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39696.12_14-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline