Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00446831_m1
          See other TRIM24 GE Assays ›
          Gene Symbol:
          Trim24
          Gene Name:
          tripartite motif-containing 24
          Gene Aliases:
          A130082H20Rik, AI447469, D430004I05Rik, TIF1, TIF1-alpha, TIF1alpha, Tif1a
          Chromosome Location:
          Chr.6: 37869880 - 37968445 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00446831_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trim24
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001272064.1 NP_001258993.1 3 - 4 929 89
          NM_001272076.1 NP_001259005.1 3 - 4 509 89
          NM_145076.4 NP_659542.3 3 - 4 929 89
          GenBank mRNA AB221637.1 - 3 - 4 629 89
          AK084870.1 - 3 - 4 921 89
          AK133144.1 - 3 - 4 509 89
          BC056959.1 - 3 - 4 907 5698079 89
          S78219.1 - 3 - 4 929 89
          S78221.1 - 3 - 4 929 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000362.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trim24

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          negative regulation of cell proliferation
          positive regulation of gene expression
          protein ubiquitination
          protein catabolic process
          regulation of protein stability
          regulation of apoptotic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          calcium ion homeostasis
          regulation of vitamin D receptor signaling pathway
          cellular response to estrogen stimulus
          regulation of signal transduction by p53 class mediator
          regulation of nucleic acid-templated transcription
          p53 binding
          DNA binding
          chromatin binding
          transcription coactivator activity
          protein kinase activity
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ligand-dependent nuclear receptor binding
          ribosomal S6-glutamic acid ligase activity
          estrogen response element binding
          sequence-specific DNA binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          ubiquitin protein ligase activity
          lysine-acetylated histone binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600002731.mm.1 TC0600000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002731.mm.1 NM_001272064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002731.mm.1 NM_001272076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002731.mm.1 NM_145076.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002735.mm.1 TC0600000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002735.mm.1 NM_001272064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002735.mm.1 NM_001272076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002735.mm.1 NM_145076.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002730.mm.1 TC0600000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002730.mm.1 NM_001272076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002733.mm.1 TC0600000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002733.mm.1 NM_001272064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002733.mm.1 NM_001272076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002733.mm.1 NM_145076.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002727.mm.1 TC0600000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002727.mm.1 NM_001272064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002727.mm.1 NM_145076.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002732.mm.1 TC0600000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002732.mm.1 NM_001272064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002732.mm.1 NM_001272076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002732.mm.1 NM_145076.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002734.mm.1 TC0600000362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600002734.mm.1 NM_001272064.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002734.mm.1 NM_001272076.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600002734.mm.1 NM_145076.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 632999 - -
          ABI Mouse Genome Survey Microarray 880915 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline