Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00447289_m1
          See other SELENBP1 GE Assays ›
          Gene Symbol:
          Selenbp1 Selenbp2
          Gene Name:
          selenium binding protein 1
          selenium binding protein 2
          Gene Aliases:
          Lp56, Lpsb, SBP56
          AP56, Lpsb2
          Chromosome Location:
          Chr.3: 94933083 - 94944758 on Build GRCm38
          Chromosome Location:
          Chr.3: 94693573 - 94704406 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00447289_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Selenbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009150.3 NP_033176.2 9 - 10 1122 103
          GenBank mRNA AK002447.1 - 9 - 10 1138 103
          BC011202.1 - 9 - 10 1122 4209393 103
          M32032.1 - 1105 103
          Gene Symbol: Selenbp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019414.2 NP_062287.1 9 - 10 103
          GenBank mRNA BC024106.1 - 9 - 10 5096831 103
          S56599.1 - 9 - 10 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000875.mm.1, TC0300000880.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Selenbp1
          Click here to find siRNAs for Gene Selenbp2

          Panther Classification:

          Molecular Function -

          defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          protein transport
          brown fat cell differentiation
          selenium binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961014 AK002447.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961014 BC011202.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961014 NM_009150.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136091 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167165 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961014 AK002447.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961014 BC011202.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961014 NM_009150.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22341356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22341348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22341333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22341339 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04993779 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00701772 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167165 - -
          Whole Mouse Genome G4122A A_52_P629487 AK002447.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629487 BC011202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629487 NM_009150.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132104 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167165 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02736828 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02736826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22341455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02567734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02736826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02913431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02736828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02770619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02913425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02540980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02682714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03073906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03150568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02682709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02540968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22341359 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494116 - -
          Affymetrix HT MG-430 PM Array Plate 1417580_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1450699_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10494114 - -
          Mouse Genome 430 2.0 Array 1450699_at - -
          Mouse Genome 430 2.0 Array 1417580_PM_s_at - -
          Mouse Genome 430 2.0 Array 1450699_PM_at - -
          Mouse Genome 430 2.0 Array 1417580_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20341_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20341_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa11680_at - -
          Affymetrix Mouse Clariom S Array PSR0300006708.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006708.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006708.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006710.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006710.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006710.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006711.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006711.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006711.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006705.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006705.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006705.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006707.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006707.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006707.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006716.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006716.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006716.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006787.mm.1 NM_009150.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006787.mm.1 NM_009150.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006787.mm.1 TC0300000880.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006698.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006698.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006698.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006713.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006713.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006713.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006786.mm.1 NM_009150.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006786.mm.1 NM_009150.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006786.mm.1 TC0300000880.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006788.mm.1 NM_009150.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006788.mm.1 NM_009150.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006788.mm.1 TC0300000880.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006706.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006706.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006706.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006712.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006712.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006712.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006699.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006699.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006699.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006700.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006700.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006700.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006715.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006715.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006715.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006783.mm.1 NM_009150.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006783.mm.1 NM_009150.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006783.mm.1 TC0300000880.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006789.mm.1 NM_009150.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006789.mm.1 NM_009150.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006789.mm.1 TC0300000880.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006694.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006694.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006694.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006701.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006701.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006701.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006703.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006703.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006703.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006714.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006714.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006714.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006709.mm.1 NM_019414.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006709.mm.1 NM_019414.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006709.mm.1 TC0300000875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006784.mm.1 NM_009150.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006784.mm.1 NM_009150.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006784.mm.1 TC0300000880.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006785.mm.1 NM_009150.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006785.mm.1 NM_009150.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006785.mm.1 TC0300000880.mm.1 MTA TC
          Mouse Expression Set 430 1417580_PM_s_at - -
          Mouse Expression Set 430 1450699_at - -
          Mouse Expression Set 430 1450699_PM_at - -
          Mouse Expression Set 430 1417580_s_at - -
          Murine Genome U74v2 Set 162030_r_at - -
          Murine Genome U74v2 Set 100596_at - -
          Murine Genome U74v2 Set 162030_PM_r_at - -
          Murine Genome U74v2 Set 100596_PM_at - -
          Murine Genome U74v2 Set 170910_r_at - -
          Murine Genome U74v2 Set 170910_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1450699_at - -
          Mouse Genome 430A 2.0 Array 1417580_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1450699_PM_at - -
          Mouse Genome 430A 2.0 Array 1417580_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 489094 - -
          ABI Mouse Genome Survey Microarray 729898 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850575 - -
          Illumina mouse-6 v1.0 expression beadchip 3850575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2727309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2742426 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2727309 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2742426 - -
          mouse-6 v1.1 (Illumina) scl020341.12_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020341.12_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2742426 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2742426 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline