Search
Search

Sept2| Gene Symbol: Sept2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159717.1 | NP_001153189.1 | 9 - 10 | 933 | 73 | |
| NM_001159718.1 | NP_001153190.1 | 8 - 9 | 906 | 73 | ||
| NM_001159719.1 | NP_001153191.1 | 9 - 10 | 981 | 73 | ||
| NM_010891.2 | NP_035021.1 | 9 - 10 | 937 | 73 | ||
| GenBank mRNA | AK028072.1 | - | 9 - 10 | 919 | 73 | |
| AK146616.1 | - | 9 - 10 | 934 | 73 | ||
| AK151591.1 | - | 9 - 10 | 921 | 73 | ||
| AK169852.1 | - | 8 - 9 | 955 | 73 | ||
| AK171331.1 | - | 9 - 10 | 916 | 73 | ||
| AK196297.1 | - | 2 - 3 | 260 | 73 | ||
| AK216929.1 | - | 2 - 3 | 260 | 73 | ||
| BC138636.1 | - | 8 - 9 | 874 | 8861656 | 73 | |
| BC138637.1 | - | 8 - 9 | 874 | 8861657 | 73 | |
| D49382.2 | - | 9 - 10 | 892 | 73 | ||
Set Membership: |
|