Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448103_m1
          See other PFDN2 GE Assays ›
          Gene Symbol:
          Pfdn2
          Gene Name:
          prefoldin 2
          Gene Aliases:
          ESTM27, W48336
          Chromosome Location:
          Chr.1: 171345699 - 171358170 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448103_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pfdn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011070.3 NP_035200.2 1 - 2 84 91
          GenBank mRNA BC026839.1 - 1 - 2 71 4219635 91
          BC049606.1 - 1 - 2 84 6535749 91
          Y17393.1 - 1 - 2 74 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001586.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pfdn2

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          positive regulation of cytoskeleton organization
          protein binding involved in protein folding
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20827418 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20827419 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 BC049606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 Y17393.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 BC026839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 NM_011070.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 BC049606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 Y17393.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 BC026839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327778 NM_011070.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 113268 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139909 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120384 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 BC049606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 Y17393.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 BC026839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 NM_011070.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 BC049606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 Y17393.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 BC026839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327778 NM_011070.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 BC049606.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 Y17393.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 BC026839.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 NM_011070.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 BC049606.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 Y17393.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 BC026839.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327778 NM_011070.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20827434 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04354143 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120384 - -
          Whole Mouse Genome G4122A A_51_P327778 BC049606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327778 Y17393.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327778 BC026839.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327778 NM_011070.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327778 BC049606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327778 Y17393.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327778 BC026839.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327778 NM_011070.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136573 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120384 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00516306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00516293 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20827419 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20827418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20827484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00394901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01055700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01055698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01055697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01055699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01070004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01070006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00394900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01055696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01085624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00603440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01070003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20827449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00560791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00650992 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10351585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10351586 - -
          Affymetrix HT MG-430 PM Array Plate 1421950_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10351583 - -
          Mouse Genome 430 2.0 Array 1421950_at - -
          Mouse Genome 430 2.0 Array 1421950_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18637_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18637_at - -
          Affymetrix Mouse Clariom S Array PSR0100013054.mm.1 NM_011070.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013054.mm.1 NM_011070.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013054.mm.1 TC0100001586.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100013056.mm.1 NM_011070.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013056.mm.1 NM_011070.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100013056.mm.1 TC0100001586.mm.1 MTA TC
          Mouse Expression Set 430 1421950_at - -
          Mouse Expression Set 430 1421950_PM_at - -
          Murine Genome U74v2 Set 102252_at - -
          Murine Genome U74v2 Set 102252_PM_at - -
          Mouse Genome 430A 2.0 Array 1421950_at - -
          Mouse Genome 430A 2.0 Array 1421950_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421950_at - -
          Affymetrix HT Mouse Genome 430A Array 1421950_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 906745 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4230156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219667 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219667 - -
          mouse-6 v1.1 (Illumina) scl018637.3_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230156 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018637.3_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219667 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219667 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018637.3_29-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline