Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448160_m1
          See other PLA2G2A GE Assays ›
          Gene Symbol:
          Pla2g2a
          Gene Name:
          phospholipase A2, group IIA (platelets, synovial fluid)
          Gene Aliases:
          EF, Mom1, Pla2, sPLA2, sPla2-IIA
          Chromosome Location:
          Chr.4: 138831857 - 138835189 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448160_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001082531.1 NP_001076000.1 4 - 5 334 105
          NR_002926.3 4 - 5 354 105
          GenBank mRNA AK131857.1 - 4 - 5 355 105
          BC045156.1 - 4 - 5 334 6479080 105
          U28244.1 - 4 - 5 341 105
          U32359.1 - 3 - 4 309 105
          X74266.1 - 4 - 5 350 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001734.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          phospholipid metabolic process
          negative regulation of cell proliferation
          lipid catabolic process
          somatic stem cell population maintenance
          regulation of growth
          regulation of cell proliferation
          negative regulation of T cell proliferation
          regulation of epithelial cell proliferation
          negative regulation of epithelial cell proliferation
          phospholipase A2 activity
          calcium ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121042 U28244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121042 U32359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121042 X74266.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121042 BC045156.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2121042 CB933893.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107224 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121042 U28244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121042 U32359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121042 X74266.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121042 BC045156.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2121042 CB933893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256632 U28244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256632 U32359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256632 X74266.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256632 BC045156.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256632 CB933893.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351255 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351246 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351240 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03351247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23348208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03351255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03351254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03351252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03203938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03774426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03351263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03771096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03351233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03351239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03351247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23348204 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509587 - -
          Affymetrix HT MG-430 PM Array Plate 1450128_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509584 - -
          Mouse Genome 430 2.0 Array 1450128_at - -
          Mouse Genome 430 2.0 Array 1450128_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18780_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18780_at - -
          Affymetrix Murine Genome 6500A Array Msa16350_at - -
          Affymetrix Mouse Clariom S Array PSR0400014164.mm.1 NR_002926.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014164.mm.1 NR_002926.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014164.mm.1 TC0400001734.mm.1 MTA TC
          Mouse Expression Set 430 1450128_PM_at - -
          Mouse Expression Set 430 1450128_at - -
          Murine Genome U74v2 Set 92735_at - -
          Murine Genome U74v2 Set 92735_PM_at - -
          Mouse Genome 430A 2.0 Array 1450128_at - -
          Mouse Genome 430A 2.0 Array 1450128_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450128_at - -
          Affymetrix HT Mouse Genome 430A Array 1450128_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 918948 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3190458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2727163 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2727163 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2727163 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline