Search
Search

Psen2| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128605.1 | NP_001122077.1 | 2 - 3 | 225 | 70 | |
| NM_011183.3 | NP_035313.2 | 2 - 3 | 234 | 70 | ||
| GenBank mRNA | AK148111.1 | - | 2 - 3 | 226 | 70 | |
| AK154119.1 | - | 2 - 3 | 170 | 70 | ||
| BC010403.1 | - | 1 - 2 | 180 | 4038761 | 70 | |
| BY136632.1 | - | 2 - 3 | 234 | 70 | ||
| U57324.1 | - | 2 - 3 | 185 | 70 | ||
Set Membership: |
|