Search
Search

Ptpn11| Gene Symbol: Ptpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109992.1 | NP_001103462.1 | 3 - 4 | 517 | 80 | |
| NM_011202.3 | NP_035332.1 | 3 - 4 | 517 | 80 | ||
| GenBank mRNA | AK011468.1 | - | 3 - 4 | 479 | 80 | |
| AK147354.1 | - | 3 - 4 | 488 | 80 | ||
| AK159501.1 | - | 3 - 4 | 517 | 80 | ||
| AK159587.1 | - | 3 - 4 | 517 | 80 | ||
| BC028513.1 | - | 3 - 4 | 467 | 80 | ||
| BC057398.1 | - | 3 - 4 | 449 | 6409640 | 80 | |
| BC059278.1 | - | 3 - 4 | 495 | 2866038 | 80 | |
| D84372.1 | - | 3 - 4 | 393 | 80 | ||
Set Membership: |
|