| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500014488.mm.1 |
NM_011293.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014488.mm.1 |
NM_011293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014488.mm.1 |
TC0500001598.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500014489.mm.1 |
NM_011293.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014489.mm.1 |
NM_011293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014489.mm.1 |
TC0500001598.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500014487.mm.1 |
NM_011293.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014487.mm.1 |
NM_011293.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500014487.mm.1 |
TC0500001598.mm.1 |
MTA TC |