Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448649_m1
          See other POLR2J GE Assays ›
          Gene Symbol:
          Polr2j
          Gene Name:
          polymerase (RNA) II (DNA directed) polypeptide J
          Gene Aliases:
          14.5kDa, Polr2i, Rpb11a, Rpo2-4
          Chromosome Location:
          Chr.5: 136116691 - 136122947 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448649_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr2j
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011293.2 NP_035423.2 2 - 3 205 68
          GenBank mRNA AK158610.1 - 2 - 3 206 68
          AK158644.1 - 2 - 3 195 68
          BC042939.1 - 2 - 3 162 6476299 68
          D85818.1 - 2 - 3 159 68
          DV065741.1 - 239 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001598.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Polr2j

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          RNA polymerase II activity
          DNA binding
          DNA-directed RNA polymerase activity
          protein binding
          LRR domain binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119518 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119513 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996643 AK167868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996643 AK158644.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996643 BC042939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996643 DV065741.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996643 NM_011293.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3707 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137728 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996643 AK167868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996643 AK158644.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996643 BC042939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996643 DV065741.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996643 NM_011293.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326086 AK158644.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326086 BC042939.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326086 D85818.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326086 DV065741.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326086 NM_011293.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24119502 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137728 - -
          Whole Mouse Genome G4122A A_51_P326086 AK158644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326086 BC042939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326086 D85818.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326086 DV065741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326086 NM_011293.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112304 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04937763 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04677941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04677939 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04677940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04677936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04937765 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04677937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119517 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24119518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24119546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04547898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04281023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04190792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04445793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04592104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04916955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04677940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04281022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04677936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24119499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526484 - -
          Affymetrix HT MG-430 PM Array Plate 1417720_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440065_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526482 - -
          Mouse Genome 430 2.0 Array 1417720_at - -
          Mouse Genome 430 2.0 Array 1417720_PM_at - -
          Mouse Genome 430 2.0 Array 1440065_PM_at - -
          Mouse Genome 430 2.0 Array 1440065_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20022_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20022_at - -
          Affymetrix Mouse Clariom S Array PSR0500014490.mm.1 TC0500001598.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500014490.mm.1 NM_011293.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500014490.mm.1 NM_011293.2 Refseq
          Mouse Expression Set 430 1417720_at - -
          Mouse Expression Set 430 1417720_PM_at - -
          Mouse Expression Set 430 1440065_PM_at - -
          Mouse Expression Set 430 1440065_at - -
          Murine Genome U74v2 Set 100461_at - -
          Murine Genome U74v2 Set 100461_PM_at - -
          Mouse Genome 430A 2.0 Array 1417720_at - -
          Mouse Genome 430A 2.0 Array 1417720_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417720_at - -
          Affymetrix HT Mouse Genome 430A Array 1417720_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440065_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440065_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 655753 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100372 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100372 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239525 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239525 - -
          mouse-6 v1.1 (Illumina) scl27119.4.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100372 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27119.4.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100372 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239525 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239525 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27119.4.1_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline