| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500029546.mm.1 |
TC0500003268.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500029546.mm.1 |
NM_001253904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029546.mm.1 |
NM_001253904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029546.mm.1 |
NM_001253905.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029546.mm.1 |
NM_001253905.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029546.mm.1 |
NM_011424.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029546.mm.1 |
NM_011424.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029547.mm.1 |
TC0500003268.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500029547.mm.1 |
NM_001253904.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029547.mm.1 |
NM_001253904.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029547.mm.1 |
NM_001253905.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029547.mm.1 |
NM_001253905.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029547.mm.1 |
NM_011424.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500029547.mm.1 |
NM_011424.3 |
Refseq |