Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448855_s1
          See other SPRR2F GE Assays ›
          Gene Symbol:
          Sprr2f
          Gene Name:
          small proline-rich protein 2F
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 92365187 - 92366442 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448855_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sprr2f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011472.2 NP_035602.1 2 396 71
          GenBank mRNA AJ005564.1 - 1 357 71
          AY158990.1 - 2 396 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000826.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sprr2f

          Gene Ontology Categories:

          Function(s) Process(es)

          epidermis development
          keratinocyte differentiation
          keratinization
          response to estradiol
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386870 AJ005564.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386870 AY158990.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386870 NM_011472.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386870 AJ005564.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386870 AY158990.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386870 NM_011472.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38901 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164101 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386870 AJ005564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386870 AY158990.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386870 NM_011472.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386870 AJ005564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386870 AY158990.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386870 NM_011472.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22331078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164101 - -
          Whole Mouse Genome G4122A A_51_P386870 AJ005564.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386870 AY158990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386870 NM_011472.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386870 AJ005564.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386870 AY158990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386870 NM_011472.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135960 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164101 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03106197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03106184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02391642 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03106186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03106175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22331101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03009513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03106181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03106195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03106182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03106183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02545424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02722984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02391648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02490745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02391641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22331101 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493871 - -
          Affymetrix HT MG-430 PM Array Plate 1449833_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493870 - -
          Mouse Genome 430 2.0 Array 1449833_at - -
          Mouse Genome 430 2.0 Array 1449833_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20760_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20760_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300006401.mm.1 NM_011472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006401.mm.1 NM_011472.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006401.mm.1 TC0300000826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006399.mm.1 NM_011472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006399.mm.1 NM_011472.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006399.mm.1 TC0300000826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300006402.mm.1 NM_011472.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006402.mm.1 NM_011472.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300006402.mm.1 TC0300000826.mm.1 MTA TC
          Mouse Expression Set 430 1449833_at - -
          Mouse Expression Set 430 1449833_PM_at - -
          Murine Genome U74v2 Set 94120_s_at - -
          Murine Genome U74v2 Set 94120_PM_s_at - -
          Murine Genome U74v2 Set 101761_f_at - -
          Murine Genome U74v2 Set 101761_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1449833_at - -
          Mouse Genome 430A 2.0 Array 1449833_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449833_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449833_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 764162 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670551 - -
          Illumina mouse-6 v1.0 expression beadchip 670551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2695944 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2695944 - -
          mouse-6 v1.1 (Illumina) scl020760.1_287-S - -
          Illumina Sentrix mouse-6.v1 bead array 670551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020760.1_287-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2695944 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2695944 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline