Search
Search
| Gene Symbol: Tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161722.1 | NP_001155194.1 | 8 - 9 | 1248 | 65 | |
| NM_001161723.1 | NP_001155195.1 | 8 - 9 | 1242 | 65 | ||
| NM_011549.3 | NP_035679.3 | 7 - 8 | 1189 | 65 | ||
| GenBank mRNA | AF079095.1 | - | 7 - 8 | 1170 | 65 | |
| AK146016.1 | - | 7 - 8 | 1214 | 65 | ||
| AK154971.1 | - | 8 - 9 | 1246 | 65 | ||
| BC064789.1 | - | 7 - 8 | 1189 | 30429545 | 65 | |
Set Membership: |
|