Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00449032_g1
          See other THBS1 GE Assays ›
          Gene Symbol:
          Thbs1
          Gene Name:
          thrombospondin 1
          Gene Aliases:
          TSP-1, TSP1, Thbs-1, tbsp1
          Chromosome Location:
          Chr.2: 118111890 - 118127133 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00449032_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thbs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011580.3 NP_035710.2 17 - 18 2953 84
          GenBank mRNA AK080686.1 - 17 - 18 2988 84
          AK145202.1 - 17 - 18 2983 84
          AK163092.1 - 17 - 18 2980 84
          AK164480.1 - 1 - 2 36 84
          AK164691.1 - 1 - 2 47 84
          BC042422.1 - 17 - 18 2955 4018176 84
          BC050917.1 - 17 - 18 2953 6405710 84
          M87276.1 - 17 - 18 2979 84
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK089315.1 - 1437 Chr. - - [-] on Build 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001729.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thbs1

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          negative regulation of endothelial cell proliferation
          negative regulation of cell-matrix adhesion
          sprouting angiogenesis
          negative regulation of antigen processing and presentation of peptide or polysaccharide antigen via MHC class II
          negative regulation of dendritic cell antigen processing and presentation
          outflow tract morphogenesis
          endocardial cushion development
          growth plate cartilage development
          inflammatory response
          response to unfolded protein
          cell cycle arrest
          cell adhesion
          response to mechanical stimulus
          response to glucose
          positive regulation of endothelial cell migration
          negative regulation of endothelial cell migration
          negative regulation of plasma membrane long-chain fatty acid transport
          negative regulation of nitric oxide mediated signal transduction
          negative regulation of cGMP-mediated signaling
          negative regulation of plasminogen activation
          positive regulation of macrophage chemotaxis
          positive regulation of fibroblast migration
          positive regulation of cell-substrate adhesion
          cell migration
          negative regulation of angiogenesis
          peptide cross-linking
          positive regulation of blood coagulation
          positive regulation of cell migration
          positive regulation of transforming growth factor beta receptor signaling pathway
          regulation of cGMP metabolic process
          response to magnesium ion
          negative regulation of interleukin-12 production
          response to endoplasmic reticulum stress
          negative regulation of fibroblast growth factor receptor signaling pathway
          positive regulation of phosphorylation
          positive regulation of tumor necrosis factor biosynthetic process
          positive regulation of macrophage activation
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of blood vessel endothelial cell migration
          negative regulation of blood vessel endothelial cell migration
          engulfment of apoptotic cell
          positive regulation of translation
          positive regulation of angiogenesis
          behavioral response to pain
          blood vessel morphogenesis
          positive regulation of chemotaxis
          response to calcium ion
          positive regulation of protein kinase B signaling
          negative regulation of fibrinolysis
          positive regulation of transforming growth factor beta production
          cellular response to nitric oxide
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of endothelial cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          negative regulation of endothelial cell chemotaxis
          phosphatidylserine binding
          fibronectin binding
          integrin binding
          calcium ion binding
          protein binding
          heparin binding
          fibroblast growth factor binding
          low-density lipoprotein particle binding
          laminin binding
          transforming growth factor beta binding
          extracellular matrix binding
          fibrinogen binding
          collagen V binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline