Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00449297_m1
          See other OAS1B GE Assays ›
          Gene Symbol:
          Oas1b
          Gene Name:
          2'-5' oligoadenylate synthetase 1B
          Gene Aliases:
          Flv, L1, Mmu-L1, Oas1, Oias-2, Oias2, Wnv
          Chromosome Location:
          Chr.5: 120812635 - 120824163 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00449297_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oas1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083925.1 NP_001077394.1 1 - 2 242 63
          NR_003507.1 1 - 2 245 63
          GenBank mRNA AB067529.1 - 1 - 2 245 63
          AF328926.1 - 1 - 2 242 63
          AF418004.1 - 1 - 2 242 63
          AF418005.1 - 1 - 2 242 63
          AF466822.1 - 1 - 2 199 63
          AK080152.1 - 1 - 2 235 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001349.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          nucleotidyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          immune response
          response to virus
          negative regulation of viral genome replication
          innate immune response
          defense response to virus
          2'-5'-oligoadenylate synthetase activity
          RNA binding
          double-stranded RNA binding
          protein binding
          ATP binding
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998962 AF418005.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998962 AF328926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998962 NM_001083925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959061 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959064 AK080152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959064 AF466822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959064 AI326028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959064 AB067529.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959064 AF418004.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 583924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50057 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959064 AK080152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959064 AF466822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959064 AI326028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959064 AB067529.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959064 AF418004.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998962 AF418005.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998962 AF328926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998962 NM_001083925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110877 AK080152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110877 AF466822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110877 AB067529.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110877 AF418004.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110877 NR_003507.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04470457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04470454 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04216361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24040998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24041072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04398584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04685524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04811941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24040999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04915756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04455463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04211755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04685522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24040998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04470456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04398586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24041030 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10525163 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525161 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525164 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525159 - -
          Affymetrix HT MG-430 PM Array Plate 1425119_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10525158 - -
          Mouse Genome 430 2.0 Array 1425119_at - -
          Mouse Genome 430 2.0 Array 1425119_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23961_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23961_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500011765.mm.1 TC0500001349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011765.mm.1 NR_003507.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011765.mm.1 NR_003507.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011762.mm.1 TC0500001349.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011762.mm.1 NR_003507.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011762.mm.1 NR_003507.1 Refseq
          Mouse Expression Set 430 1425119_PM_at - -
          Mouse Expression Set 430 1425119_at - -
          Murine Genome U74v2 Set 137038_at - -
          Murine Genome U74v2 Set 137038_PM_at - -
          Mouse Genome 430A 2.0 Array 1425119_at - -
          Mouse Genome 430A 2.0 Array 1425119_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425119_at - -
          Affymetrix HT Mouse Genome 430A Array 1425119_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 667080 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2260402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613140 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613140 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2823462 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2613140 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613140 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2823462 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline