Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00449560_m1
          See other PNCK GE Assays ›
          Gene Symbol:
          Pnck
          Gene Name:
          pregnancy upregulated non-ubiquitously expressed CaM kinase
          Gene Aliases:
          Bstk3, CaMKIbeta2, CaMKlb2, Camk1b, Punc, caMKlb1
          Chromosome Location:
          Chr.X: 73655992 - 73660127 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00449560_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pnck
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199351.1 NP_001186280.1 2 - 3 253 73
          NM_001199352.1 NP_001186281.1 2 - 3 117 73
          NM_012040.3 NP_036170.1 2 - 3 193 73
          GenBank mRNA AB023027.1 - 1 - 2 63 73
          AF181984.1 - 2 - 3 167 73
          AK134665.1 - 2 - 3 124 73
          AK158755.1 - 2 - 3 118 73
          BC051996.1 - 2 - 3 100 73
          BC055891.1 - 2 - 3 93 4208888 73
          CX243772.1 - 2 - 3 253 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002431.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pnck

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phosphorylation
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          calmodulin binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32464005 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32464004 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969615 AK158755.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969615 BC051996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969615 BC055891.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969615 CD776443.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969615 NM_001199352.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254672 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127375 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969615 AK158755.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969615 BC051996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969615 BC055891.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969615 CD776443.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969615 NM_001199352.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270324 AK158755.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270324 BC051996.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270324 BC055891.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270324 NM_001199352.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P270324 NM_012040.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32463987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32464023 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127375 - -
          Whole Mouse Genome G4122A A_51_P270324 AK158755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270324 BC051996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270324 BC055891.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270324 NM_001199352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P270324 NM_012040.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102857 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109633 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127375 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14612493 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14815301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14815303 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14815308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14612489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14815296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32464005 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32464004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32464025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32463966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15157713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14612493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15157705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15157685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14803844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15157707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14596518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15157702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14813855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14815307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14815301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14596514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32463985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32464018 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605072 - -
          Affymetrix HT MG-430 PM Array Plate 1422711_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10605067 - -
          Mouse Genome 430 2.0 Array 1422711_a_at - -
          Mouse Genome 430 2.0 Array 1422711_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93843_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93843_at - -
          Affymetrix Mouse Clariom S Array PSR0X00015047.mm.1 NM_001199352.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015047.mm.1 NM_001199352.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015047.mm.1 TC0X00002431.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00015040.mm.1 NM_001199351.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015040.mm.1 NM_001199351.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015040.mm.1 NM_001199352.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015040.mm.1 NM_001199352.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015040.mm.1 NM_012040.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015040.mm.1 NM_012040.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00015040.mm.1 TC0X00002431.mm.1 MTA TC
          Mouse Expression Set 430 1422711_PM_a_at - -
          Mouse Expression Set 430 1422711_a_at - -
          Murine Genome U74v2 Set 93910_at - -
          Murine Genome U74v2 Set 93910_PM_at - -
          Mouse Genome 430A 2.0 Array 1422711_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422711_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422711_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422711_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 320725 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6040538 - -
          Illumina mouse-6 v1.0 expression beadchip 6040538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040538 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244364 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244364 - -
          mouse-6 v1.1 (Illumina) scl54159.12.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54159.12.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244364 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244364 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl54159.12.1_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline