Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00450613_m1
          See other SULT1C1 GE Assays ›
          Gene Symbol:
          Sult1c1
          Gene Name:
          sulfotransferase family, cytosolic, 1C, member 1
          Gene Aliases:
          (PST)G, P-SULT, ST1C1, Stp2, Sult1a2, mOLFST
          Chromosome Location:
          Chr.17: 53961615 - 53990631 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00450613_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sult1c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018751.2 NP_061221.2 3 - 4 438 71
          GenBank mRNA AF033653.1 - 3 - 4 449 71
          BC045149.1 - 3 - 4 438 6308175 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002292.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sult1c1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          sulfur compound metabolic process
          metabolic process
          N-acetylglucosamine 6-O-sulfotransferase activity
          N-acetylgalactosamine 4-O-sulfotransferase activity
          alcohol sulfotransferase activity
          aryl sulfotransferase activity
          heparan sulfate 2-O-sulfotransferase activity
          sulfotransferase activity
          HNK-1 sulfotransferase activity
          transferase activity
          heparan sulfate 6-O-sulfotransferase activity
          trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
          1-phenanthrol sulfotransferase activity
          3-phenanthrol sulfotransferase activity
          4-phenanthrol sulfotransferase activity
          trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
          9-phenanthrol sulfotransferase activity
          2-phenanthrol sulfotransferase activity
          phenanthrol sulfotransferase activity
          1-hydroxypyrene sulfotransferase activity
          galactose 3-O-sulfotransferase activity
          proteoglycan sulfotransferase activity
          cholesterol sulfotransferase activity
          hydroxyjasmonate sulfotransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987740 BC045149.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1987740 NM_018751.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135387 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121639 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987740 BC045149.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1987740 NM_018751.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P519811 AF033653.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31245950 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08806922 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08806875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135387 - -
          Whole Mouse Genome G4122A A_51_P519811 AF033653.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112617 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121639 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135387 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125862 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125855 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31245929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13422733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13459441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13180246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13302028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13144708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13317079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13335794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13422724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13422732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31245936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13180245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451837 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451831 - -
          Affymetrix HT MG-430 PM Array Plate 1420470_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451828 - -
          Mouse Genome 430 2.0 Array 1420470_at - -
          Mouse Genome 430 2.0 Array 1420470_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20888_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20888_at - -
          Affymetrix Mouse Clariom S Array PSR1700021434.mm.1 NM_018751.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021434.mm.1 NM_018751.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021434.mm.1 TC1700002292.mm.1 MTA TC
          Mouse Expression Set 430 1420470_PM_at - -
          Mouse Expression Set 430 1420470_at - -
          Murine Genome U74v2 Set 96421_PM_at - -
          Murine Genome U74v2 Set 96421_at - -
          Mouse Genome 430A 2.0 Array 1420470_at - -
          Mouse Genome 430A 2.0 Array 1420470_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 585754 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1740348 - -
          Illumina mouse-6 v1.0 expression beadchip 1740348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2713826 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2713826 - -
          mouse-6 v1.1 (Illumina) scl49792.8.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49792.8.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2713826 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2713826 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline