| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22343685 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22343686 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22343684 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965635 |
BC060173.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965635 |
BC079537.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965635 |
AK122198.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965635 |
AK088590.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965635 |
AK132202.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
302224 |
AK139600.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
302224 |
AK162136.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
302224 |
NM_207261.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906189 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
695942 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
607134 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
166407 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674422 |
AF017369.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674422 |
AK051395.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674422 |
AK157835.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674422 |
AK165413.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674422 |
BC132340.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
674422 |
NM_015759.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
767572 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
377074 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
204700 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
299968 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
208738 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
93963 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758661 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932506 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574421 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
182684 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
909065 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
166218 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
682617 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
908414 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
51058 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
292549 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574876 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
919459 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
30151 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
6346 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
172307 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
18679 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P163015 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P104908 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965635 |
BC060173.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965635 |
BC079537.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965635 |
AK122198.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965635 |
AK088590.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965635 |
AK132202.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P609475 |
BC007176.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P609475 |
BC060173.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P609475 |
BC079537.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P609475 |
AK122198.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P609475 |
AK088590.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P31717 |
AF546078.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343703 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343701 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343684 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343704 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343689 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343687 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343686 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343698 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22343667 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04994878 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04994928 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00702482 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P163015 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04994844 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P104908 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P609475 |
BC007176.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P609475 |
BC060173.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P609475 |
BC079537.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P609475 |
AK122198.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P609475 |
AK088590.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P31717 |
AF546078.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105770 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P104908 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P163015 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04994928 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02431141 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02888906 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03138895 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22343686 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22343684 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22343685 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343551 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343563 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343624 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343533 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343491 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343548 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343450 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343580 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343656 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343585 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343502 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343493 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343516 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343595 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343686 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343593 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343539 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343511 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343451 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343514 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343475 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343513 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343495 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343636 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343472 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343621 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343441 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343550 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343474 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343570 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343515 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343497 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343481 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343559 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343535 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343468 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343505 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343588 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343433 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343484 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343564 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343609 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343575 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343569 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343592 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343437 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343447 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343689 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343465 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343602 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343704 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343541 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343473 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343632 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343494 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343467 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343530 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343536 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343478 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343480 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343618 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343503 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343486 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343556 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343577 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343499 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343526 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343508 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343462 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343701 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343457 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343584 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343612 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343507 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343561 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343614 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343620 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343434 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343456 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343611 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343524 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343445 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343642 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343487 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343440 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343496 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343509 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343628 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343517 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343470 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343459 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343594 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343652 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343439 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343504 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343637 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22343698 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02810989 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343567 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343533 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03154882 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343559 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02888906 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343704 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343698 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02645994 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343650 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03106945 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343520 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02355637 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343644 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02791225 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343610 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343509 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343659 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343654 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343705 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02453521 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343687 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343689 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02997259 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02360380 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343706 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343561 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03106950 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343543 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343536 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02888915 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343516 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343521 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343656 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02756039 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343524 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343555 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343566 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343511 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343514 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343529 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343612 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03106946 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03156646 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343563 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02711840 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343701 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343647 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343564 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02810980 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343645 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343526 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02627212 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343541 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343539 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22343652 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03013536 |
- |
- |